Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 132624740-132624965 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POLE
Entrez Gene ID
5426
Gene Name
DNA polymerase epsilon, catalytic subunit
Gene Aliases
CRCS12, FILS, POLE1
UniGene
Hs.524871
Gene Chromosome Location
Chr.12: 132623762-132687524 on Build GRCh38
Transcripts
NM_006231.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6752048 c.6715G>A p.A2239T Chr.12: 132624937-132624937 on Build GRCh38
6978625 c.6743C>T p.T2248I Chr.12: 132624909-132624909 on Build GRCh38
937251 c.6714C>T p.C2238C Chr.12: 132624938-132624938 on Build GRCh38
7093188 c.6726C>T p.F2242F Chr.12: 132624926-132624926 on Build GRCh38
6929311 c.6806A>G p.Y2269C Chr.12: 132624752-132624752 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768405124 Chr.12: 132624774-132624774 on Build GRCh38
rs5745082 Chr.12: 132624855-132624859 on Build GRCh38
rs775513803 Chr.12: 132624876-132624876 on Build GRCh38
rs774472240 Chr.12: 132624941-132624941 on Build GRCh38
rs199947622 Chr.12: 132624856-132624859 on Build GRCh38
rs759660507 Chr.12: 132624804-132624804 on Build GRCh38
rs917917396 Chr.12: 132624920-132624920 on Build GRCh38
rs761616241 Chr.12: 132624857-132624866 on Build GRCh38
rs376336585 Chr.12: 132624793-132624793 on Build GRCh38
rs371596124 Chr.12: 132624847-132624847 on Build GRCh38
rs771312122 Chr.12: 132624926-132624926 on Build GRCh38
rs200911338 Chr.12: 132624762-132624762 on Build GRCh38
rs773619926 Chr.12: 132624850-132624850 on Build GRCh38
rs548289800 Chr.12: 132624917-132624917 on Build GRCh38
rs139246810 Chr.12: 132624830-132624830 on Build GRCh38
rs750255126 Chr.12: 132624816-132624816 on Build GRCh38
rs749707316 Chr.12: 132624791-132624791 on Build GRCh38
rs748851961 Chr.12: 132624764-132624764 on Build GRCh38
rs531461991 Chr.12: 132624882-132624882 on Build GRCh38
rs148604472 Chr.12: 132624855-132624863 on Build GRCh38
rs753711213 Chr.12: 132624856-132624864 on Build GRCh38
rs747852529 Chr.12: 132624845-132624845 on Build GRCh38
rs750363700 Chr.12: 132624853-132624854 on Build GRCh38
rs757147672 Chr.12: 132624862-132624870 on Build GRCh38
rs747349009 Chr.12: 132624925-132624925 on Build GRCh38
rs142222159 Chr.12: 132624763-132624763 on Build GRCh38
rs866548835 Chr.12: 132624783-132624783 on Build GRCh38
rs755999901 Chr.12: 132624883-132624883 on Build GRCh38
rs200082120 Chr.12: 132624938-132624938 on Build GRCh38
rs71452531 Chr.12: 132624785-132624785 on Build GRCh38
rs763221440 Chr.12: 132624877-132624877 on Build GRCh38
rs375041812 Chr.12: 132624862-132624866 on Build GRCh38
rs200926632 Chr.12: 132624866-132624866 on Build GRCh38
rs777145520 Chr.12: 132624863-132624863 on Build GRCh38
rs1009675656 Chr.12: 132624839-132624839 on Build GRCh38
rs373768478 Chr.12: 132624807-132624807 on Build GRCh38
rs375741031 Chr.12: 132624922-132624922 on Build GRCh38
rs780192918 Chr.12: 132624829-132624829 on Build GRCh38
rs540203276 Chr.12: 132624781-132624781 on Build GRCh38
rs764265575 Chr.12: 132624881-132624881 on Build GRCh38
rs754481643 Chr.12: 132624908-132624908 on Build GRCh38
rs116323660 Chr.12: 132624792-132624792 on Build GRCh38
rs756931710 Chr.12: 132624810-132624810 on Build GRCh38
rs777051903 Chr.12: 132624933-132624933 on Build GRCh38
rs879057349 Chr.12: 132624820-132624820 on Build GRCh38
rs753832634 Chr.12: 132624899-132624899 on Build GRCh38
rs1026222953 Chr.12: 132624828-132624828 on Build GRCh38
rs376140352 Chr.12: 132624801-132624801 on Build GRCh38
rs777338519 Chr.12: 132624924-132624924 on Build GRCh38
rs552311764 Chr.12: 132624821-132624821 on Build GRCh38
rs73155056 Chr.12: 132624795-132624795 on Build GRCh38
rs78686967 Chr.12: 132624857-132624857 on Build GRCh38
rs568888626 Chr.12: 132624863-132624866 on Build GRCh38
rs762044635 Chr.12: 132624782-132624782 on Build GRCh38
rs770341236 Chr.12: 132624935-132624935 on Build GRCh38
rs767356249 Chr.12: 132624814-132624814 on Build GRCh38
rs771404326 Chr.12: 132624858-132624858 on Build GRCh38
rs369576300 Chr.12: 132624803-132624803 on Build GRCh38
rs747676884 Chr.12: 132624918-132624918 on Build GRCh38
rs765301412 Chr.12: 132624869-132624869 on Build GRCh38
rs766379073 Chr.12: 132624889-132624889 on Build GRCh38
rs190813054 Chr.12: 132624936-132624936 on Build GRCh38
rs550706326 Chr.12: 132624857-132624861 on Build GRCh38
rs370305500 Chr.12: 132624937-132624937 on Build GRCh38
rs748762369 Chr.12: 132624833-132624833 on Build GRCh38
rs746384198 Chr.12: 132624934-132624934 on Build GRCh38
rs376902172 Chr.12: 132624838-132624838 on Build GRCh38

View Full Product Details