Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 211360985-211361250 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRAF5
Entrez Gene ID
7188
Gene Name
TNF receptor associated factor 5
Gene Aliases
MGC:39780, RNF84
UniGene
Hs.523930
Gene Chromosome Location
Chr.1: 211326587-211374946 on Build GRCh38
Transcripts
NM_001033910.2 NM_001319207.1 NM_004619.3 NM_145759.2 XM_011509957.2 XM_011509959.2 XM_011509960.2 XM_017002221.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1338620 c.695C>T p.T232M Chr.1: 211361161-211361161 on Build GRCh38
6045352 c.680A>G p.Y227C Chr.1: 211361146-211361146 on Build GRCh38
7252571 c.696+9T>C p.? Chr.1: 211361171-211361171 on Build GRCh38
5646160 c.649G>A p.E217K Chr.1: 211361115-211361115 on Build GRCh38
1165395 c.667C>G p.P223A Chr.1: 211361133-211361133 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs760253888 Chr.1: 211361063-211361063 on Build GRCh38
rs750647287 Chr.1: 211361194-211361194 on Build GRCh38
rs757526659 Chr.1: 211361084-211361084 on Build GRCh38
rs939437158 Chr.1: 211361079-211361079 on Build GRCh38
rs747904561 Chr.1: 211361162-211361162 on Build GRCh38
rs929150324 Chr.1: 211361009-211361009 on Build GRCh38
rs753735879 Chr.1: 211361078-211361078 on Build GRCh38
rs769735226 Chr.1: 211361208-211361208 on Build GRCh38
rs1018784646 Chr.1: 211361023-211361023 on Build GRCh38
rs771914729 Chr.1: 211361137-211361137 on Build GRCh38
rs1038640496 Chr.1: 211361201-211361201 on Build GRCh38
rs763548566 Chr.1: 211361175-211361176 on Build GRCh38
rs756288732 Chr.1: 211361199-211361199 on Build GRCh38
rs540788952 Chr.1: 211361092-211361092 on Build GRCh38
rs746269758 Chr.1: 211361187-211361187 on Build GRCh38
rs766478189 Chr.1: 211361069-211361069 on Build GRCh38
rs199606735 Chr.1: 211361083-211361083 on Build GRCh38
rs748490218 Chr.1: 211361131-211361131 on Build GRCh38
rs767253629 Chr.1: 211361046-211361046 on Build GRCh38
rs79833851 Chr.1: 211361153-211361153 on Build GRCh38
rs113925874 Chr.1: 211361161-211361161 on Build GRCh38
rs772982474 Chr.1: 211361146-211361146 on Build GRCh38
rs769155761 Chr.1: 211361114-211361114 on Build GRCh38
rs145765066 Chr.1: 211361147-211361147 on Build GRCh38
rs370068812 Chr.1: 211361110-211361110 on Build GRCh38
rs752477069 Chr.1: 211361188-211361188 on Build GRCh38
rs967615617 Chr.1: 211361026-211361026 on Build GRCh38
rs377012367 Chr.1: 211361104-211361104 on Build GRCh38
rs781363454 Chr.1: 211361089-211361089 on Build GRCh38
rs754882722 Chr.1: 211361080-211361080 on Build GRCh38
rs1008694405 Chr.1: 211361008-211361008 on Build GRCh38
rs908742975 Chr.1: 211361173-211361173 on Build GRCh38
rs774626101 Chr.1: 211361123-211361123 on Build GRCh38
rs780018024 Chr.1: 211361211-211361211 on Build GRCh38
rs1051703147 Chr.1: 211361216-211361216 on Build GRCh38
rs561538704 Chr.1: 211361006-211361006 on Build GRCh38
rs374585053 Chr.1: 211361157-211361157 on Build GRCh38
rs140223127 Chr.1: 211361038-211361038 on Build GRCh38
rs765202118 Chr.1: 211361178-211361178 on Build GRCh38
rs200289585 Chr.1: 211361103-211361103 on Build GRCh38
rs1018766682 Chr.1: 211361007-211361007 on Build GRCh38
rs370725672 Chr.1: 211361062-211361062 on Build GRCh38
rs36091380 Chr.1: 211361025-211361026 on Build GRCh38

View Full Product Details