Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 18489805-18490072 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBBP9
Entrez Gene ID
10741
Gene Name
RB binding protein 9, serine hydrolase
Gene Aliases
BOG, RBBP10
UniGene
Hs.69330
Gene Chromosome Location
Chr.20: 18486540-18497250 on Build GRCh38
Transcripts
NM_006606.2 XM_005260652.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3933794 c.343A>C p.T115P Chr.20: 18489982-18489982 on Build GRCh38
723515 c.411C>T p.D137D Chr.20: 18489914-18489914 on Build GRCh38
6456396 c.491G>A p.G164D Chr.20: 18489834-18489834 on Build GRCh38
6413203 c.442G>T p.E148* Chr.20: 18489883-18489883 on Build GRCh38
3389611 c.417G>A p.P139P Chr.20: 18489908-18489908 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754652462 Chr.20: 18489870-18489870 on Build GRCh38
rs953784165 Chr.20: 18490034-18490034 on Build GRCh38
rs145407527 Chr.20: 18489979-18489979 on Build GRCh38
rs149743101 Chr.20: 18489904-18489904 on Build GRCh38
rs151179936 Chr.20: 18489858-18489858 on Build GRCh38
rs6111990 Chr.20: 18489953-18489953 on Build GRCh38
rs868056849 Chr.20: 18489964-18489964 on Build GRCh38
rs781227980 Chr.20: 18489995-18489995 on Build GRCh38
rs761137904 Chr.20: 18489949-18489949 on Build GRCh38
rs778200197 Chr.20: 18489883-18489883 on Build GRCh38
rs756708853 Chr.20: 18489998-18489999 on Build GRCh38
rs748295433 Chr.20: 18489999-18489999 on Build GRCh38
rs773193704 Chr.20: 18490008-18490008 on Build GRCh38
rs565219087 Chr.20: 18489914-18489914 on Build GRCh38
rs1013166608 Chr.20: 18489977-18489977 on Build GRCh38
rs3748450 Chr.20: 18490020-18490020 on Build GRCh38
rs200476717 Chr.20: 18489964-18489964 on Build GRCh38
rs147934779 Chr.20: 18489888-18489888 on Build GRCh38
rs774945907 Chr.20: 18489975-18489975 on Build GRCh38
rs368159380 Chr.20: 18489978-18489978 on Build GRCh38
rs746932582 Chr.20: 18489900-18489900 on Build GRCh38
rs774369398 Chr.20: 18489913-18489913 on Build GRCh38
rs560343448 Chr.20: 18489874-18489874 on Build GRCh38
rs3748449 Chr.20: 18490002-18490002 on Build GRCh38
rs201470200 Chr.20: 18489879-18489879 on Build GRCh38
rs1027159370 Chr.20: 18490023-18490023 on Build GRCh38
rs986052718 Chr.20: 18490044-18490044 on Build GRCh38
rs751533283 Chr.20: 18489999-18489999 on Build GRCh38
rs772030006 Chr.20: 18489920-18489920 on Build GRCh38
rs774883241 Chr.20: 18489960-18489960 on Build GRCh38
rs145102973 Chr.20: 18489886-18489886 on Build GRCh38
rs773815963 Chr.20: 18489937-18489937 on Build GRCh38
rs973800426 Chr.20: 18489859-18489859 on Build GRCh38
rs781104713 Chr.20: 18489908-18489908 on Build GRCh38
rs199550613 Chr.20: 18489994-18489994 on Build GRCh38
rs376202865 Chr.20: 18489933-18489933 on Build GRCh38
rs745619409 Chr.20: 18489909-18489909 on Build GRCh38
rs916282308 Chr.20: 18489894-18489894 on Build GRCh38
rs367676775 Chr.20: 18489959-18489959 on Build GRCh38
rs62216369 Chr.20: 18489875-18489875 on Build GRCh38
rs149149837 Chr.20: 18489980-18489980 on Build GRCh38
rs777059995 Chr.20: 18490032-18490032 on Build GRCh38
rs960651454 Chr.20: 18490038-18490038 on Build GRCh38
rs780822033 Chr.20: 18490007-18490009 on Build GRCh38
rs923611053 Chr.20: 18489982-18489982 on Build GRCh38
rs746539455 Chr.20: 18490021-18490021 on Build GRCh38
rs779251417 Chr.20: 18489997-18489997 on Build GRCh38
rs147115402 Chr.20: 18489967-18489967 on Build GRCh38
rs745662321 Chr.20: 18489996-18489996 on Build GRCh38

View Full Product Details