Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 38209568-38209813 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SSTR1
Entrez Gene ID
6751
Gene Name
somatostatin receptor 1
Gene Aliases
SRIF-2, SS-1-R, SS1-R, SS1R
UniGene
Hs.248160
Gene Chromosome Location
Chr.14: 38207999-38213063 on Build GRCh38
Transcripts
NM_001049.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7211667 c.290T>C p.I97T Chr.14: 38209679-38209679 on Build GRCh38
6320392 c.360G>T p.L120F Chr.14: 38209749-38209749 on Build GRCh38
5007281 c.389G>A p.C130Y Chr.14: 38209778-38209778 on Build GRCh38
270245 c.409G>A p.D137N Chr.14: 38209798-38209798 on Build GRCh38
1664358 c.418A>C p.N140H Chr.14: 38209807-38209807 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751004108 Chr.14: 38209592-38209592 on Build GRCh38
rs755985842 Chr.14: 38209633-38209633 on Build GRCh38
rs759340668 Chr.14: 38209688-38209688 on Build GRCh38
rs773093431 Chr.14: 38209716-38209716 on Build GRCh38
rs757226690 Chr.14: 38209663-38209663 on Build GRCh38
rs146928458 Chr.14: 38209632-38209632 on Build GRCh38
rs747079308 Chr.14: 38209666-38209666 on Build GRCh38
rs1018834855 Chr.14: 38209670-38209675 on Build GRCh38
rs768744926 Chr.14: 38209686-38209686 on Build GRCh38
rs753890091 Chr.14: 38209661-38209661 on Build GRCh38
rs773561220 Chr.14: 38209765-38209765 on Build GRCh38
rs904387798 Chr.14: 38209712-38209712 on Build GRCh38
rs766994626 Chr.14: 38209791-38209791 on Build GRCh38
rs951635346 Chr.14: 38209705-38209705 on Build GRCh38
rs774880721 Chr.14: 38209785-38209785 on Build GRCh38
rs746645070 Chr.14: 38209782-38209782 on Build GRCh38
rs759283314 Chr.14: 38209593-38209593 on Build GRCh38
rs771406317 Chr.14: 38209777-38209777 on Build GRCh38
rs891169932 Chr.14: 38209638-38209638 on Build GRCh38
rs770202303 Chr.14: 38209751-38209751 on Build GRCh38
rs774199997 Chr.14: 38209653-38209655 on Build GRCh38
rs981480635 Chr.14: 38209767-38209767 on Build GRCh38
rs763493236 Chr.14: 38209773-38209773 on Build GRCh38
rs1025850614 Chr.14: 38209682-38209682 on Build GRCh38
rs995667216 Chr.14: 38209678-38209678 on Build GRCh38
rs962832105 Chr.14: 38209673-38209673 on Build GRCh38
rs1042903870 Chr.14: 38209662-38209662 on Build GRCh38
rs777696699 Chr.14: 38209649-38209649 on Build GRCh38
rs781277793 Chr.14: 38209719-38209719 on Build GRCh38
rs748315729 Chr.14: 38209741-38209741 on Build GRCh38
rs767105630 Chr.14: 38209621-38209621 on Build GRCh38

View Full Product Details