Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 47631553-47631824 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTCH2
Entrez Gene ID
23788
Gene Name
mitochondrial carrier 2
Gene Aliases
HSPC032, MIMP, SLC25A50
UniGene
Hs.269944
Gene Chromosome Location
Chr.11: 47604309-47642654 on Build GRCh38
Transcripts
NM_001317231.1 NM_001317232.1 NM_001317233.1 NM_014342.3 XM_006718172.2 XM_011519959.2 XM_011519960.2 XM_011519961.2 XM_017017462.1 XM_017017463.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
927469 c.392G>A p.R131H Chr.11: 47631689-47631689 on Build GRCh38
6716187 c.404C>T p.T135I Chr.11: 47631677-47631677 on Build GRCh38
4033230 c.388G>A p.A130T Chr.11: 47631693-47631693 on Build GRCh38
3447853 c.391C>T p.R131C Chr.11: 47631690-47631690 on Build GRCh38
6132679 c.388G>T p.A130S Chr.11: 47631693-47631693 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201001139 Chr.11: 47631758-47631758 on Build GRCh38
rs999896333 Chr.11: 47631663-47631663 on Build GRCh38
rs761945742 Chr.11: 47631763-47631763 on Build GRCh38
rs371558162 Chr.11: 47631716-47631716 on Build GRCh38
rs1056501977 Chr.11: 47631593-47631593 on Build GRCh38
rs377150975 Chr.11: 47631719-47631719 on Build GRCh38
rs534200387 Chr.11: 47631630-47631630 on Build GRCh38
rs773289740 Chr.11: 47631747-47631747 on Build GRCh38
rs373590179 Chr.11: 47631635-47631635 on Build GRCh38
rs530285224 Chr.11: 47631751-47631751 on Build GRCh38
rs769138128 Chr.11: 47631738-47631739 on Build GRCh38
rs954177479 Chr.11: 47631791-47631791 on Build GRCh38
rs762745340 Chr.11: 47631751-47631751 on Build GRCh38
rs761871292 Chr.11: 47631627-47631627 on Build GRCh38
rs368256862 Chr.11: 47631720-47631720 on Build GRCh38
rs758792658 Chr.11: 47631708-47631708 on Build GRCh38
rs752980443 Chr.11: 47631676-47631676 on Build GRCh38
rs768223805 Chr.11: 47631620-47631620 on Build GRCh38
rs766025055 Chr.11: 47631648-47631648 on Build GRCh38
rs377203241 Chr.11: 47631731-47631731 on Build GRCh38
rs764267048 Chr.11: 47631686-47631686 on Build GRCh38
rs763342157 Chr.11: 47631605-47631605 on Build GRCh38
rs765300969 Chr.11: 47631629-47631629 on Build GRCh38
rs776229768 Chr.11: 47631625-47631625 on Build GRCh38
rs753960764 Chr.11: 47631689-47631689 on Build GRCh38
rs774693054 Chr.11: 47631743-47631744 on Build GRCh38
rs746073691 Chr.11: 47631767-47631767 on Build GRCh38
rs201087705 Chr.11: 47631693-47631693 on Build GRCh38
rs374980101 Chr.11: 47631792-47631792 on Build GRCh38
rs749881528 Chr.11: 47631784-47631784 on Build GRCh38
rs1018801571 Chr.11: 47631714-47631714 on Build GRCh38
rs756245198 Chr.11: 47631679-47631679 on Build GRCh38
rs779455823 Chr.11: 47631699-47631699 on Build GRCh38
rs896049969 Chr.11: 47631581-47631581 on Build GRCh38
rs775279851 Chr.11: 47631608-47631608 on Build GRCh38
rs754047176 Chr.11: 47631761-47631761 on Build GRCh38
rs371023319 Chr.11: 47631771-47631771 on Build GRCh38
rs1044674641 Chr.11: 47631584-47631584 on Build GRCh38
rs950288891 Chr.11: 47631583-47631583 on Build GRCh38
rs746574483 Chr.11: 47631613-47631613 on Build GRCh38
rs144753500 Chr.11: 47631704-47631704 on Build GRCh38
rs541447446 Chr.11: 47631694-47631694 on Build GRCh38
rs766222261 Chr.11: 47631752-47631752 on Build GRCh38
rs747707008 Chr.11: 47631727-47631727 on Build GRCh38
rs772310094 Chr.11: 47631605-47631605 on Build GRCh38
rs796924967 Chr.11: 47631690-47631690 on Build GRCh38
rs368031896 Chr.11: 47631641-47631641 on Build GRCh38
rs764351632 Chr.11: 47631760-47631760 on Build GRCh38
rs909920975 Chr.11: 47631793-47631793 on Build GRCh38
rs138711289 Chr.11: 47631580-47631580 on Build GRCh38
rs139957971 Chr.11: 47631709-47631709 on Build GRCh38

View Full Product Details