Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 172925455-172925728 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPATA16
Entrez Gene ID
83893
Gene Name
spermatogenesis associated 16
Gene Aliases
NYD-SP12, SPGF6
UniGene
Hs.444236
Gene Chromosome Location
Chr.3: 172889357-173141268 on Build GRCh38
Transcripts
NM_031955.5 XM_006713778.2 XM_017007308.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4931402 c.1083T>A p.D361E Chr.3: 172925491-172925491 on Build GRCh38
1617218 c.1110G>A p.Q370Q Chr.3: 172925464-172925464 on Build GRCh38
7133474 c.1087C>G p.Q363E Chr.3: 172925487-172925487 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs779445099 Chr.3: 172925508-172925508 on Build GRCh38
rs528721959 Chr.3: 172925692-172925692 on Build GRCh38
rs146509162 Chr.3: 172925510-172925510 on Build GRCh38
rs905927369 Chr.3: 172925675-172925675 on Build GRCh38
rs370476094 Chr.3: 172925522-172925522 on Build GRCh38
rs763512486 Chr.3: 172925580-172925580 on Build GRCh38
rs1053405598 Chr.3: 172925560-172925560 on Build GRCh38
rs934906978 Chr.3: 172925556-172925556 on Build GRCh38
rs367603647 Chr.3: 172925551-172925551 on Build GRCh38
rs757986283 Chr.3: 172925503-172925503 on Build GRCh38
rs902185583 Chr.3: 172925554-172925554 on Build GRCh38
rs748564972 Chr.3: 172925523-172925523 on Build GRCh38
rs981652308 Chr.3: 172925505-172925505 on Build GRCh38
rs895957592 Chr.3: 172925571-172925571 on Build GRCh38
rs1023105466 Chr.3: 172925665-172925665 on Build GRCh38
rs994889955 Chr.3: 172925678-172925678 on Build GRCh38
rs373835470 Chr.3: 172925531-172925531 on Build GRCh38
rs769872637 Chr.3: 172925530-172925530 on Build GRCh38
rs749896332 Chr.3: 172925484-172925484 on Build GRCh38
rs34376487 Chr.3: 172925546-172925546 on Build GRCh38
rs755513313 Chr.3: 172925521-172925521 on Build GRCh38
rs899483617 Chr.3: 172925589-172925589 on Build GRCh38
rs753784960 Chr.3: 172925532-172925533 on Build GRCh38
rs757136436 Chr.3: 172925533-172925534 on Build GRCh38
rs139979680 Chr.3: 172925549-172925549 on Build GRCh38
rs749313389 Chr.3: 172925539-172925539 on Build GRCh38
rs998143454 Chr.3: 172925613-172925613 on Build GRCh38

View Full Product Details