Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 102924858-102925109 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAM
Entrez Gene ID
5066
Gene Name
peptidylglycine alpha-amidating monooxygenase
Gene Aliases
PAL, PHM
UniGene
Hs.369430
Gene Chromosome Location
Chr.5: 102754783-103031105 on Build GRCh38
Transcripts
NM_000919.3 NM_001177306.1 NM_001319943.1 NM_138766.2 NM_138821.2 NM_138822.2 NR_033440.1 XM_011543419.2 XM_017009495.1 XM_017009496.1 XM_017009497.1 XM_017009498.1 XM_017009499.1 XM_017009500.1 XM_017009501.1 XM_017009502.1 XM_017009503.1 XM_017009504.1 XM_017009505.1 XM_017009506.1 XM_017009507.1 XM_017009508.1 XM_017009509.1 XM_017009510.1 XM_017009511.1 XM_017009512.1 XM_017009513.1 XM_017009514.1 XM_017009515.1 XM_017009516.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6483181 c.408G>T p.W136C Chr.5: 102925008-102925008 on Build GRCh38
4979620 c.427A>T p.T143S Chr.5: 102925027-102925027 on Build GRCh38
1059252 c.413G>T p.R138I Chr.5: 102925013-102925013 on Build GRCh38
4788697 c.396T>G p.I132M Chr.5: 102924996-102924996 on Build GRCh38
6483182 c.408G>T p.W136C Chr.5: 102925008-102925008 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201178915 Chr.5: 102924993-102924993 on Build GRCh38
rs778364854 Chr.5: 102925030-102925030 on Build GRCh38
rs779600645 Chr.5: 102924916-102924916 on Build GRCh38
rs374960531 Chr.5: 102925077-102925077 on Build GRCh38
rs746916354 Chr.5: 102925057-102925057 on Build GRCh38
rs772958107 Chr.5: 102924893-102924893 on Build GRCh38
rs377205864 Chr.5: 102924999-102924999 on Build GRCh38
rs764426860 Chr.5: 102924995-102924995 on Build GRCh38
rs754777893 Chr.5: 102924924-102924924 on Build GRCh38
rs771728520 Chr.5: 102924881-102924881 on Build GRCh38
rs748938597 Chr.5: 102924922-102924922 on Build GRCh38
rs749543332 Chr.5: 102925071-102925071 on Build GRCh38
rs375939839 Chr.5: 102924905-102924905 on Build GRCh38
rs931060307 Chr.5: 102924986-102924986 on Build GRCh38
rs200199885 Chr.5: 102925067-102925067 on Build GRCh38
rs775617960 Chr.5: 102924980-102924980 on Build GRCh38
rs762105269 Chr.5: 102925005-102925005 on Build GRCh38
rs765889557 Chr.5: 102925020-102925020 on Build GRCh38
rs748161430 Chr.5: 102924938-102924938 on Build GRCh38
rs370166108 Chr.5: 102925011-102925011 on Build GRCh38
rs991163138 Chr.5: 102924940-102924940 on Build GRCh38
rs750886218 Chr.5: 102925015-102925015 on Build GRCh38
rs777884799 Chr.5: 102925040-102925040 on Build GRCh38
rs755658446 Chr.5: 102925017-102925017 on Build GRCh38
rs972457576 Chr.5: 102924964-102924964 on Build GRCh38
rs745967580 Chr.5: 102924945-102924945 on Build GRCh38
rs181340861 Chr.5: 102925031-102925031 on Build GRCh38
rs754653876 Chr.5: 102925026-102925026 on Build GRCh38
rs769878323 Chr.5: 102924970-102924970 on Build GRCh38
rs769817582 Chr.5: 102925060-102925060 on Build GRCh38
rs768032395 Chr.5: 102925010-102925010 on Build GRCh38
rs753491201 Chr.5: 102925021-102925021 on Build GRCh38
rs772141514 Chr.5: 102924939-102924939 on Build GRCh38
rs372778602 Chr.5: 102924942-102924942 on Build GRCh38
rs371195536 Chr.5: 102924930-102924930 on Build GRCh38
rs755568652 Chr.5: 102924908-102924908 on Build GRCh38

View Full Product Details