Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 103558695-103558966 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUDT12
Entrez Gene ID
83594
Gene Name
nudix hydrolase 12
Gene Aliases
-
UniGene
Hs.434289
Gene Chromosome Location
Chr.5: 103548855-103562805 on Build GRCh38
Transcripts
NM_001300741.1 NM_031438.3 XM_005272095.1 XM_005272097.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1059322 c.737A>C p.N246T Chr.5: 103558938-103558938 on Build GRCh38
7429018 c.733G>A p.E245K Chr.5: 103558942-103558942 on Build GRCh38
2990079 c.794C>A p.A265D Chr.5: 103558881-103558881 on Build GRCh38
7281025 c.775C>T p.Q259* Chr.5: 103558900-103558900 on Build GRCh38
4895888 c.743A>T p.Y248F Chr.5: 103558932-103558932 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs536962766 Chr.5: 103558814-103558814 on Build GRCh38
rs752332829 Chr.5: 103558932-103558932 on Build GRCh38
rs776937188 Chr.5: 103558914-103558914 on Build GRCh38
rs533704799 Chr.5: 103558804-103558804 on Build GRCh38
rs935682640 Chr.5: 103558744-103558744 on Build GRCh38
rs1007589723 Chr.5: 103558938-103558938 on Build GRCh38
rs953285969 Chr.5: 103558884-103558884 on Build GRCh38
rs745689177 Chr.5: 103558867-103558867 on Build GRCh38
rs746962238 Chr.5: 103558828-103558828 on Build GRCh38
rs748770936 Chr.5: 103558883-103558886 on Build GRCh38
rs977190118 Chr.5: 103558729-103558729 on Build GRCh38
rs761226452 Chr.5: 103558908-103558908 on Build GRCh38
rs143222967 Chr.5: 103558927-103558927 on Build GRCh38
rs780991749 Chr.5: 103558864-103558864 on Build GRCh38
rs925676287 Chr.5: 103558739-103558739 on Build GRCh38
rs894767376 Chr.5: 103558754-103558754 on Build GRCh38
rs770813786 Chr.5: 103558831-103558831 on Build GRCh38
rs531048945 Chr.5: 103558795-103558795 on Build GRCh38
rs773209905 Chr.5: 103558899-103558899 on Build GRCh38
rs777130273 Chr.5: 103558822-103558822 on Build GRCh38
rs766813521 Chr.5: 103558912-103558912 on Build GRCh38
rs1045948429 Chr.5: 103558792-103558792 on Build GRCh38
rs772190248 Chr.5: 103558898-103558898 on Build GRCh38
rs758051342 Chr.5: 103558936-103558936 on Build GRCh38
rs551134382 Chr.5: 103558801-103558801 on Build GRCh38
rs944053134 Chr.5: 103558916-103558916 on Build GRCh38
rs993602548 Chr.5: 103558812-103558812 on Build GRCh38
rs762186408 Chr.5: 103558889-103558889 on Build GRCh38
rs768935736 Chr.5: 103558869-103558869 on Build GRCh38
rs760123292 Chr.5: 103558855-103558855 on Build GRCh38
rs139871589 Chr.5: 103558926-103558926 on Build GRCh38
rs781568433 Chr.5: 103558872-103558874 on Build GRCh38
rs759895624 Chr.5: 103558915-103558915 on Build GRCh38
rs950352488 Chr.5: 103558781-103558781 on Build GRCh38
rs547275129 Chr.5: 103558881-103558881 on Build GRCh38
rs3734087 Chr.5: 103558746-103558746 on Build GRCh38
rs559455312 Chr.5: 103558878-103558878 on Build GRCh38

View Full Product Details