Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 5059250-5059511 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR52E2
Entrez Gene ID
119678
Gene Name
olfactory receptor family 52 subfamily E member 2
Gene Aliases
-
UniGene
Hs.553557
Gene Chromosome Location
Chr.11: 5058650-5059627 on Build GRCh38
Transcripts
NM_001005164.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1218734 c.164G>A p.S55N Chr.11: 5059464-5059464 on Build GRCh38
6054593 c.229A>T p.T77S Chr.11: 5059399-5059399 on Build GRCh38
79008 c.264G>A p.W88* Chr.11: 5059364-5059364 on Build GRCh38
5360959 c.261C>T p.F87F Chr.11: 5059367-5059367 on Build GRCh38
4819030 c.192C>T p.F64F Chr.11: 5059436-5059436 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150524739 Chr.11: 5059348-5059348 on Build GRCh38
rs757816644 Chr.11: 5059411-5059411 on Build GRCh38
rs529497286 Chr.11: 5059375-5059375 on Build GRCh38
rs180948995 Chr.11: 5059324-5059324 on Build GRCh38
rs767727689 Chr.11: 5059381-5059381 on Build GRCh38
rs201001738 Chr.11: 5059346-5059346 on Build GRCh38
rs764500547 Chr.11: 5059318-5059318 on Build GRCh38
rs752365259 Chr.11: 5059349-5059349 on Build GRCh38
rs1033740519 Chr.11: 5059380-5059380 on Build GRCh38
rs199936023 Chr.11: 5059431-5059431 on Build GRCh38
rs112235231 Chr.11: 5059351-5059351 on Build GRCh38
rs749130512 Chr.11: 5059448-5059448 on Build GRCh38
rs756573992 Chr.11: 5059304-5059304 on Build GRCh38
rs780988361 Chr.11: 5059424-5059424 on Build GRCh38
rs956279720 Chr.11: 5059398-5059398 on Build GRCh38
rs775753526 Chr.11: 5059387-5059387 on Build GRCh38
rs550773411 Chr.11: 5059401-5059401 on Build GRCh38
rs911545221 Chr.11: 5059456-5059456 on Build GRCh38
rs201714539 Chr.11: 5059335-5059335 on Build GRCh38
rs769187663 Chr.11: 5059436-5059436 on Build GRCh38
rs766528791 Chr.11: 5059287-5059287 on Build GRCh38
rs758055315 Chr.11: 5059468-5059488 on Build GRCh38
rs368842488 Chr.11: 5059337-5059337 on Build GRCh38
rs139521750 Chr.11: 5059422-5059422 on Build GRCh38
rs369925067 Chr.11: 5059379-5059379 on Build GRCh38
rs754272704 Chr.11: 5059403-5059403 on Build GRCh38
rs373112219 Chr.11: 5059389-5059389 on Build GRCh38
rs546284517 Chr.11: 5059325-5059325 on Build GRCh38
rs765941352 Chr.11: 5059420-5059420 on Build GRCh38
rs1004634666 Chr.11: 5059274-5059274 on Build GRCh38
rs749286745 Chr.11: 5059358-5059358 on Build GRCh38
rs564526108 Chr.11: 5059334-5059334 on Build GRCh38
rs144077173 Chr.11: 5059293-5059293 on Build GRCh38
rs944354951 Chr.11: 5059464-5059464 on Build GRCh38
rs12576484 Chr.11: 5059361-5059361 on Build GRCh38
rs757828384 Chr.11: 5059319-5059319 on Build GRCh38
rs747868316 Chr.11: 5059428-5059428 on Build GRCh38
rs774524681 Chr.11: 5059289-5059289 on Build GRCh38
rs113958818 Chr.11: 5059423-5059423 on Build GRCh38
rs145335319 Chr.11: 5059453-5059453 on Build GRCh38
rs374783754 Chr.11: 5059297-5059297 on Build GRCh38
rs774345486 Chr.11: 5059454-5059454 on Build GRCh38
rs1035870303 Chr.11: 5059397-5059397 on Build GRCh38
rs772194905 Chr.11: 5059485-5059485 on Build GRCh38

View Full Product Details