Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 69796729-69797000 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC10A1
Entrez Gene ID
6554
Gene Name
solute carrier family 10 member 1
Gene Aliases
NTCP
UniGene
Hs.952
Gene Chromosome Location
Chr.14: 69775835-69797289 on Build GRCh38
Transcripts
NM_003049.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
344314 c.265G>C p.E89Q Chr.14: 69796891-69796891 on Build GRCh38
1225841 c.200C>T p.A67V Chr.14: 69796956-69796956 on Build GRCh38
2135793 c.202C>T p.Q68* Chr.14: 69796954-69796954 on Build GRCh38
120021 c.280T>G p.L94V Chr.14: 69796876-69796876 on Build GRCh38
4767204 c.247T>G p.F83V Chr.14: 69796909-69796909 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1050593691 Chr.14: 69796873-69796873 on Build GRCh38
rs777187143 Chr.14: 69796857-69796857 on Build GRCh38
rs761020846 Chr.14: 69796831-69796831 on Build GRCh38
rs4646285 Chr.14: 69796931-69796931 on Build GRCh38
rs201086542 Chr.14: 69796791-69796791 on Build GRCh38
rs917082692 Chr.14: 69796787-69796787 on Build GRCh38
rs138880008 Chr.14: 69796813-69796813 on Build GRCh38
rs772287060 Chr.14: 69796910-69796910 on Build GRCh38
rs745783074 Chr.14: 69796808-69796808 on Build GRCh38
rs370448356 Chr.14: 69796789-69796789 on Build GRCh38
rs1001561751 Chr.14: 69796847-69796847 on Build GRCh38
rs201366799 Chr.14: 69796906-69796906 on Build GRCh38
rs117354704 Chr.14: 69796772-69796772 on Build GRCh38
rs750853881 Chr.14: 69796821-69796821 on Build GRCh38
rs767858944 Chr.14: 69796820-69796820 on Build GRCh38
rs769833293 Chr.14: 69796811-69796811 on Build GRCh38
rs147401668 Chr.14: 69796863-69796863 on Build GRCh38
rs368178722 Chr.14: 69796898-69796898 on Build GRCh38
rs745418645 Chr.14: 69796906-69796906 on Build GRCh38
rs201979229 Chr.14: 69796918-69796918 on Build GRCh38
rs761126161 Chr.14: 69796919-69796919 on Build GRCh38
rs963911045 Chr.14: 69796916-69796916 on Build GRCh38
rs199728494 Chr.14: 69796805-69796805 on Build GRCh38
rs148467625 Chr.14: 69796893-69796893 on Build GRCh38
rs770817085 Chr.14: 69796794-69796794 on Build GRCh38
rs774528826 Chr.14: 69796816-69796816 on Build GRCh38
rs752257139 Chr.14: 69796779-69796779 on Build GRCh38
rs202018997 Chr.14: 69796966-69796966 on Build GRCh38
rs756925388 Chr.14: 69796760-69796760 on Build GRCh38
rs778283161 Chr.14: 69796773-69796773 on Build GRCh38
rs201764278 Chr.14: 69796874-69796874 on Build GRCh38
rs771644365 Chr.14: 69796936-69796936 on Build GRCh38
rs778901974 Chr.14: 69796970-69796970 on Build GRCh38
rs763683476 Chr.14: 69796939-69796939 on Build GRCh38
rs200816244 Chr.14: 69796868-69796868 on Build GRCh38
rs150649857 Chr.14: 69796932-69796932 on Build GRCh38
rs755908953 Chr.14: 69796962-69796962 on Build GRCh38
rs748244007 Chr.14: 69796971-69796971 on Build GRCh38
rs899258109 Chr.14: 69796861-69796861 on Build GRCh38
rs781181196 Chr.14: 69796798-69796798 on Build GRCh38
rs778455372 Chr.14: 69796796-69796807 on Build GRCh38
rs757955614 Chr.14: 69796851-69796851 on Build GRCh38
rs757932511 Chr.14: 69796788-69796788 on Build GRCh38
rs767290491 Chr.14: 69796950-69796950 on Build GRCh38
rs940899684 Chr.14: 69796953-69796953 on Build GRCh38
rs201712905 Chr.14: 69796899-69796899 on Build GRCh38
rs779764405 Chr.14: 69796965-69796965 on Build GRCh38
rs200957618 Chr.14: 69796839-69796839 on Build GRCh38
rs768763066 Chr.14: 69796895-69796895 on Build GRCh38
rs201078426 Chr.14: 69796967-69796967 on Build GRCh38
rs201800239 Chr.14: 69796937-69796937 on Build GRCh38
rs750134733 Chr.14: 69796961-69796961 on Build GRCh38
rs201638287 Chr.14: 69796841-69796841 on Build GRCh38
rs895937650 Chr.14: 69796870-69796870 on Build GRCh38
rs200105842 Chr.14: 69796879-69796879 on Build GRCh38
rs200366757 Chr.14: 69796941-69796941 on Build GRCh38
rs751180988 Chr.14: 69796940-69796940 on Build GRCh38
rs760009351 Chr.14: 69796935-69796935 on Build GRCh38
rs142898958 Chr.14: 69796860-69796860 on Build GRCh38
rs756902429 Chr.14: 69796949-69796949 on Build GRCh38
rs201048210 Chr.14: 69796956-69796956 on Build GRCh38
rs200060447 Chr.14: 69796905-69796905 on Build GRCh38

View Full Product Details