Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 74338674-74338947 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLIP2
Entrez Gene ID
7461
Gene Name
CAP-Gly domain containing linker protein 2
Gene Aliases
CLIP, CLIP-115, CYLN2, WBSCR3, WBSCR4, WSCR3, WSCR4
UniGene
Hs.647018
Gene Chromosome Location
Chr.7: 74289437-74405943 on Build GRCh38
Transcripts
NM_003388.4 NM_032421.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4544115 c.349G>A p.V117M Chr.7: 74338675-74338675 on Build GRCh38
5407873 c.374G>A p.G125D Chr.7: 74338700-74338700 on Build GRCh38
3785766 c.424C>G p.P142A Chr.7: 74338750-74338750 on Build GRCh38
5795211 c.563G>A p.R188Q Chr.7: 74338889-74338889 on Build GRCh38
4162423 c.594T>C p.T198T Chr.7: 74338920-74338920 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782462347 Chr.7: 74338894-74338894 on Build GRCh38
rs372682888 Chr.7: 74338746-74338746 on Build GRCh38
rs1033608959 Chr.7: 74338764-74338764 on Build GRCh38
rs781933004 Chr.7: 74338776-74338776 on Build GRCh38
rs970551590 Chr.7: 74338707-74338707 on Build GRCh38
rs782704562 Chr.7: 74338845-74338845 on Build GRCh38
rs782228688 Chr.7: 74338765-74338765 on Build GRCh38
rs782770325 Chr.7: 74338701-74338701 on Build GRCh38
rs782204827 Chr.7: 74338823-74338823 on Build GRCh38
rs782055620 Chr.7: 74338862-74338862 on Build GRCh38
rs539518 Chr.7: 74338920-74338920 on Build GRCh38
rs782685051 Chr.7: 74338730-74338730 on Build GRCh38
rs782755384 Chr.7: 74338904-74338904 on Build GRCh38
rs782080153 Chr.7: 74338791-74338791 on Build GRCh38
rs142064745 Chr.7: 74338806-74338806 on Build GRCh38
rs782755044 Chr.7: 74338881-74338881 on Build GRCh38
rs201883164 Chr.7: 74338814-74338814 on Build GRCh38
rs199904386 Chr.7: 74338745-74338745 on Build GRCh38
rs142629570 Chr.7: 74338831-74338831 on Build GRCh38
rs532111952 Chr.7: 74338922-74338922 on Build GRCh38
rs75967845 Chr.7: 74338897-74338897 on Build GRCh38
rs112969082 Chr.7: 74338695-74338695 on Build GRCh38
rs111609508 Chr.7: 74338803-74338803 on Build GRCh38
rs782603609 Chr.7: 74338911-74338911 on Build GRCh38
rs200921421 Chr.7: 74338710-74338710 on Build GRCh38
rs781911130 Chr.7: 74338812-74338812 on Build GRCh38
rs200574175 Chr.7: 74338771-74338771 on Build GRCh38
rs782005016 Chr.7: 74338788-74338788 on Build GRCh38
rs61739991 Chr.7: 74338861-74338861 on Build GRCh38
rs782756175 Chr.7: 74338780-74338780 on Build GRCh38
rs782326408 Chr.7: 74338866-74338866 on Build GRCh38
rs782634260 Chr.7: 74338815-74338815 on Build GRCh38
rs782346420 Chr.7: 74338857-74338857 on Build GRCh38
rs782801599 Chr.7: 74338807-74338807 on Build GRCh38
rs782197995 Chr.7: 74338837-74338837 on Build GRCh38
rs782174816 Chr.7: 74338871-74338871 on Build GRCh38
rs782264415 Chr.7: 74338852-74338852 on Build GRCh38
rs782543322 Chr.7: 74338729-74338729 on Build GRCh38
rs782075860 Chr.7: 74338878-74338878 on Build GRCh38
rs782204931 Chr.7: 74338725-74338725 on Build GRCh38
rs373921467 Chr.7: 74338773-74338773 on Build GRCh38
rs368911321 Chr.7: 74338724-74338724 on Build GRCh38
rs782520628 Chr.7: 74338900-74338900 on Build GRCh38
rs978929785 Chr.7: 74338761-74338761 on Build GRCh38
rs781787750 Chr.7: 74338891-74338891 on Build GRCh38
rs138503584 Chr.7: 74338874-74338874 on Build GRCh38
rs782229360 Chr.7: 74338912-74338912 on Build GRCh38
rs2301972 Chr.7: 74338692-74338692 on Build GRCh38
rs781944542 Chr.7: 74338759-74338759 on Build GRCh38
rs201687475 Chr.7: 74338876-74338876 on Build GRCh38
rs543387438 Chr.7: 74338805-74338805 on Build GRCh38
rs782148023 Chr.7: 74338779-74338779 on Build GRCh38
rs781823126 Chr.7: 74338704-74338704 on Build GRCh38
rs782027919 Chr.7: 74338868-74338868 on Build GRCh38
rs782405366 Chr.7: 74338844-74338844 on Build GRCh38
rs782667586 Chr.7: 74338895-74338895 on Build GRCh38
rs782754682 Chr.7: 74338872-74338872 on Build GRCh38

View Full Product Details