Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 18584188-18584435 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDKL5
Entrez Gene ID
6792
Gene Name
cyclin dependent kinase like 5
Gene Aliases
CFAP247, EIEE2, ISSX, STK9
UniGene
Hs.659851
Gene Chromosome Location
Chr.X: 18425605-18653629 on Build GRCh38
Transcripts
NM_001037343.1 NM_001323289.1 NM_003159.2

SNP Detail

SNP ID Chromosome Location
rs587783111 Chr.X: 18584348-18584351 on Build GRCh38
rs764376734 Chr.X: 18584236-18584236 on Build GRCh38
rs786204988 Chr.X: 18584310-18584311 on Build GRCh38
rs369535258 Chr.X: 18584333-18584333 on Build GRCh38
rs786204986 Chr.X: 18584262-18584262 on Build GRCh38
rs786204989 Chr.X: 18584327-18584327 on Build GRCh38
rs911879541 Chr.X: 18584372-18584372 on Build GRCh38
rs267608481 Chr.X: 18584223-18584226 on Build GRCh38
rs747862334 Chr.X: 18584390-18584390 on Build GRCh38
rs863225066 Chr.X: 18584269-18584269 on Build GRCh38
rs755385076 Chr.X: 18584363-18584363 on Build GRCh38
rs267608497 Chr.X: 18584348-18584349 on Build GRCh38
rs1015920438 Chr.X: 18584302-18584302 on Build GRCh38
rs786204987 Chr.X: 18584305-18584306 on Build GRCh38
rs184524881 Chr.X: 18584224-18584224 on Build GRCh38
rs587783084 Chr.X: 18584325-18584325 on Build GRCh38
rs757402424 Chr.X: 18584272-18584272 on Build GRCh38
rs372854990 Chr.X: 18584404-18584404 on Build GRCh38
rs750878642 Chr.X: 18584279-18584279 on Build GRCh38
rs794727912 Chr.X: 18584340-18584340 on Build GRCh38
rs61749704 Chr.X: 18584338-18584338 on Build GRCh38
rs267608490 Chr.X: 18584312-18584312 on Build GRCh38
rs374030184 Chr.X: 18584254-18584254 on Build GRCh38
rs776099061 Chr.X: 18584250-18584250 on Build GRCh38
rs587783085 Chr.X: 18584341-18584341 on Build GRCh38
rs765303813 Chr.X: 18584276-18584276 on Build GRCh38
rs758844150 Chr.X: 18584285-18584285 on Build GRCh38
rs61749700 Chr.X: 18584324-18584324 on Build GRCh38
rs267608493 Chr.X: 18584331-18584331 on Build GRCh38
rs267606715 Chr.X: 18584332-18584332 on Build GRCh38
rs267608498 Chr.X: 18584364-18584364 on Build GRCh38
rs797044858 Chr.X: 18584313-18584313 on Build GRCh38
rs769192115 Chr.X: 18584209-18584209 on Build GRCh38
rs267608480 Chr.X: 18584261-18584261 on Build GRCh38

View Full Product Details