Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1637367-1637602 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK11B
Entrez Gene ID
984
Gene Name
cyclin dependent kinase 11B
Gene Aliases
CDC2L1, CDK11, CDK11-p110, CDK11-p46, CDK11-p58, CLK-1, PITSLREA, PK58, p58, p58CDC2L1, p58CLK-1
UniGene
Hs.651228
Gene Chromosome Location
Chr.1: 1635226-1659839 on Build GRCh38
Transcripts
NM_001291345.1 NM_001787.2 NM_033486.2 NM_033487.2 NM_033489.2 NM_033490.2 XM_006711065.3 XM_011542490.2 XM_011542491.2 XM_011542492.2 XM_011542493.2 XM_011542494.2 XM_011542495.2 XM_011542496.2 XM_017002925.1 XM_017002926.1 XM_017002927.1 XM_017002928.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4223164 c.1528G>A p.D510N Chr.1: 1637453-1637453 on Build GRCh38
5074449 c.1518T>C p.Y506Y Chr.1: 1637463-1637463 on Build GRCh38
3849294 c.1579+10C>T p.? Chr.1: 1637398-1637398 on Build GRCh38
1985051 c.1534G>A p.D512N Chr.1: 1637453-1637453 on Build GRCh38
6560869 c.1554G>A p.M518I Chr.1: 1637427-1637427 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs562467865 Chr.1: 1637457-1637457 on Build GRCh38
rs753334765 Chr.1: 1637454-1637454 on Build GRCh38
rs747259510 Chr.1: 1637477-1637477 on Build GRCh38
rs377296900 Chr.1: 1637524-1637524 on Build GRCh38
rs778183683 Chr.1: 1637475-1637475 on Build GRCh38
rs760252594 Chr.1: 1637513-1637513 on Build GRCh38
rs770743271 Chr.1: 1637563-1637563 on Build GRCh38
rs370329622 Chr.1: 1637464-1637464 on Build GRCh38
rs577826090 Chr.1: 1637392-1637392 on Build GRCh38
rs564859169 Chr.1: 1637410-1637410 on Build GRCh38
rs118121437 Chr.1: 1637573-1637573 on Build GRCh38
rs746060304 Chr.1: 1637540-1637540 on Build GRCh38
rs759152916 Chr.1: 1637520-1637520 on Build GRCh38
rs776307625 Chr.1: 1637519-1637519 on Build GRCh38
rs748103392 Chr.1: 1637549-1637549 on Build GRCh38
rs781262255 Chr.1: 1637490-1637490 on Build GRCh38
rs755620889 Chr.1: 1637398-1637398 on Build GRCh38
rs543963349 Chr.1: 1637415-1637415 on Build GRCh38
rs772365906 Chr.1: 1637402-1637402 on Build GRCh38
rs372517313 Chr.1: 1637404-1637404 on Build GRCh38
rs761539305 Chr.1: 1637390-1637390 on Build GRCh38
rs767313483 Chr.1: 1637435-1637435 on Build GRCh38
rs776685455 Chr.1: 1637421-1637421 on Build GRCh38
rs28688376 Chr.1: 1637577-1637577 on Build GRCh38
rs376482852 Chr.1: 1637582-1637582 on Build GRCh38
rs754457183 Chr.1: 1637399-1637399 on Build GRCh38
rs374328905 Chr.1: 1637516-1637516 on Build GRCh38
rs750093758 Chr.1: 1637439-1637439 on Build GRCh38
rs547721306 Chr.1: 1637569-1637569 on Build GRCh38
rs768521706 Chr.1: 1637424-1637424 on Build GRCh38
rs751287754 Chr.1: 1637389-1637389 on Build GRCh38
rs774153370 Chr.1: 1637430-1637430 on Build GRCh38
rs557982944 Chr.1: 1637388-1637388 on Build GRCh38
rs756235826 Chr.1: 1637545-1637545 on Build GRCh38
rs758937144 Chr.1: 1637528-1637528 on Build GRCh38
rs748469960 Chr.1: 1637401-1637401 on Build GRCh38
rs757431996 Chr.1: 1637487-1637487 on Build GRCh38
rs766056795 Chr.1: 1637451-1637451 on Build GRCh38
rs762863154 Chr.1: 1637387-1637387 on Build GRCh38
rs764367893 Chr.1: 1637531-1637531 on Build GRCh38
rs757491699 Chr.1: 1637534-1637534 on Build GRCh38
rs751891745 Chr.1: 1637532-1637532 on Build GRCh38
rs541550497 Chr.1: 1637508-1637508 on Build GRCh38
rs527238552 Chr.1: 1637554-1637554 on Build GRCh38
rs753220778 Chr.1: 1637474-1637474 on Build GRCh38
rs370740219 Chr.1: 1637525-1637525 on Build GRCh38
rs963159388 Chr.1: 1637553-1637553 on Build GRCh38
rs375978633 Chr.1: 1637463-1637463 on Build GRCh38
rs760363996 Chr.1: 1637443-1637443 on Build GRCh38
rs745898300 Chr.1: 1637491-1637491 on Build GRCh38
rs746773824 Chr.1: 1637556-1637556 on Build GRCh38
rs202208701 Chr.1: 1637426-1637426 on Build GRCh38
rs747228446 Chr.1: 1637412-1637412 on Build GRCh38
rs376521780 Chr.1: 1637400-1637400 on Build GRCh38
rs954579028 Chr.1: 1637574-1637574 on Build GRCh38
rs768611687 Chr.1: 1637492-1637492 on Build GRCh38
rs771981481 Chr.1: 1637552-1637552 on Build GRCh38
rs780095789 Chr.1: 1637548-1637548 on Build GRCh38
rs774312968 Chr.1: 1637502-1637502 on Build GRCh38
rs764814264 Chr.1: 1637521-1637521 on Build GRCh38
rs773057444 Chr.1: 1637509-1637509 on Build GRCh38

View Full Product Details