Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 28434373-28434632 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZSCAN23
Entrez Gene ID
222696
Gene Name
zinc finger and SCAN domain containing 23
Gene Aliases
ZNF390, ZNF453, dJ29K1.3, dJ29K1.3.1
UniGene
Hs.721841
Gene Chromosome Location
Chr.6: 28425516-28443554 on Build GRCh38
Transcripts
NM_001012455.1 XM_005248950.2 XM_011514406.2 XM_011514408.2 XM_011514410.2 XM_017010479.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3249993 c.1093G>T p.E365* Chr.6: 28434542-28434542 on Build GRCh38
5542513 c.1157C>T p.A386V Chr.6: 28434478-28434478 on Build GRCh38
3994787 c.1067C>T p.T356I Chr.6: 28434568-28434568 on Build GRCh38
1077126 c.1058G>T p.R353I Chr.6: 28434577-28434577 on Build GRCh38
3249992 c.1100G>C p.G367A Chr.6: 28434535-28434535 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs61730236 Chr.6: 28434604-28434604 on Build GRCh38
rs760196095 Chr.6: 28434479-28434479 on Build GRCh38
rs185399222 Chr.6: 28434583-28434583 on Build GRCh38
rs1007418856 Chr.6: 28434503-28434503 on Build GRCh38
rs771386325 Chr.6: 28434605-28434605 on Build GRCh38
rs759246782 Chr.6: 28434497-28434497 on Build GRCh38
rs74762071 Chr.6: 28434477-28434477 on Build GRCh38
rs769641401 Chr.6: 28434560-28434560 on Build GRCh38
rs267600925 Chr.6: 28434476-28434476 on Build GRCh38
rs771940239 Chr.6: 28434445-28434445 on Build GRCh38
rs764531252 Chr.6: 28434561-28434561 on Build GRCh38
rs757416478 Chr.6: 28434587-28434587 on Build GRCh38
rs746724226 Chr.6: 28434442-28434442 on Build GRCh38
rs1021750151 Chr.6: 28434495-28434495 on Build GRCh38
rs145918786 Chr.6: 28434496-28434496 on Build GRCh38
rs765633728 Chr.6: 28434553-28434553 on Build GRCh38
rs1005657284 Chr.6: 28434598-28434598 on Build GRCh38
rs17315648 Chr.6: 28434483-28434483 on Build GRCh38
rs750658989 Chr.6: 28434602-28434602 on Build GRCh38
rs1041295550 Chr.6: 28434484-28434484 on Build GRCh38
rs192872804 Chr.6: 28434558-28434558 on Build GRCh38
rs756355910 Chr.6: 28434603-28434603 on Build GRCh38
rs888094109 Chr.6: 28434593-28434593 on Build GRCh38
rs1043128721 Chr.6: 28434449-28434449 on Build GRCh38
rs780573981 Chr.6: 28434509-28434509 on Build GRCh38
rs781507382 Chr.6: 28434601-28434601 on Build GRCh38
rs569835258 Chr.6: 28434422-28434422 on Build GRCh38
rs753045218 Chr.6: 28434556-28434556 on Build GRCh38

View Full Product Details