Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 76079612-76079853 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZACN
Entrez Gene ID
353174
Gene Name
zinc activated ion channel
Gene Aliases
L2, LGICZ, LGICZ1, ZAC, ZAC1
UniGene
Hs.714919
Gene Chromosome Location
Chr.17: 76079182-76082804 on Build GRCh38
Transcripts
NM_180990.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5031956 c.232C>G p.R78G Chr.17: 76079711-76079711 on Build GRCh38
3522469 c.248C>T p.S83F Chr.17: 76079727-76079727 on Build GRCh38
4069799 c.232C>T p.R78* Chr.17: 76079711-76079711 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs781491291 Chr.17: 76079724-76079724 on Build GRCh38
rs767150915 Chr.17: 76079766-76079766 on Build GRCh38
rs373901649 Chr.17: 76079650-76079650 on Build GRCh38
rs760152946 Chr.17: 76079797-76079797 on Build GRCh38
rs903605986 Chr.17: 76079826-76079826 on Build GRCh38
rs752134812 Chr.17: 76079699-76079699 on Build GRCh38
rs114867401 Chr.17: 76079793-76079793 on Build GRCh38
rs369660049 Chr.17: 76079651-76079651 on Build GRCh38
rs764054967 Chr.17: 76079798-76079798 on Build GRCh38
rs773825209 Chr.17: 76079730-76079730 on Build GRCh38
rs777300001 Chr.17: 76079695-76079695 on Build GRCh38
rs778725073 Chr.17: 76079712-76079712 on Build GRCh38
rs756108910 Chr.17: 76079701-76079701 on Build GRCh38
rs759113634 Chr.17: 76079764-76079764 on Build GRCh38
rs779784382 Chr.17: 76079720-76079728 on Build GRCh38
rs760047265 Chr.17: 76079802-76079802 on Build GRCh38
rs772392608 Chr.17: 76079654-76079654 on Build GRCh38
rs997149808 Chr.17: 76079747-76079747 on Build GRCh38
rs370157774 Chr.17: 76079666-76079666 on Build GRCh38
rs373419607 Chr.17: 76079663-76079663 on Build GRCh38
rs892028711 Chr.17: 76079792-76079792 on Build GRCh38
rs377543076 Chr.17: 76079665-76079665 on Build GRCh38
rs550251960 Chr.17: 76079721-76079721 on Build GRCh38
rs377430797 Chr.17: 76079690-76079690 on Build GRCh38
rs999147289 Chr.17: 76079675-76079675 on Build GRCh38
rs773772239 Chr.17: 76079683-76079683 on Build GRCh38
rs774719253 Chr.17: 76079757-76079757 on Build GRCh38
rs770422618 Chr.17: 76079678-76079678 on Build GRCh38
rs763477841 Chr.17: 76079744-76079744 on Build GRCh38
rs140302218 Chr.17: 76079711-76079711 on Build GRCh38
rs570259408 Chr.17: 76079726-76079726 on Build GRCh38
rs893001065 Chr.17: 76079697-76079697 on Build GRCh38
rs567890838 Chr.17: 76079694-76079694 on Build GRCh38
rs770473701 Chr.17: 76079729-76079729 on Build GRCh38
rs536879025 Chr.17: 76079706-76079706 on Build GRCh38
rs548207801 Chr.17: 76079667-76079667 on Build GRCh38
rs766096852 Chr.17: 76079689-76079689 on Build GRCh38
rs775550156 Chr.17: 76079660-76079660 on Build GRCh38
rs142902667 Chr.17: 76079751-76079751 on Build GRCh38
rs374179633 Chr.17: 76079687-76079687 on Build GRCh38

View Full Product Details