Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 68231849-68232110 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESRP2
Entrez Gene ID
80004
Gene Name
epithelial splicing regulatory protein 2
Gene Aliases
RBM35B
UniGene
Hs.436585
Gene Chromosome Location
Chr.16: 68228547-68236233 on Build GRCh38
Transcripts
NM_024939.2 XM_005256153.4 XM_006721273.3 XM_011523338.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6265861 c.1053C>A p.F351L Chr.16: 68232078-68232078 on Build GRCh38
1609550 c.1181A>T p.D394V Chr.16: 68231920-68231920 on Build GRCh38
6656465 c.1194C>A p.L398L Chr.16: 68231907-68231907 on Build GRCh38
1379086 c.1073C>T p.S358L Chr.16: 68232028-68232028 on Build GRCh38
972708 c.1135G>A p.E379K Chr.16: 68231966-68231966 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774450234 Chr.16: 68231912-68231912 on Build GRCh38
rs772241122 Chr.16: 68232082-68232082 on Build GRCh38
rs756576950 Chr.16: 68232063-68232063 on Build GRCh38
rs201685431 Chr.16: 68231873-68231873 on Build GRCh38
rs1033623250 Chr.16: 68232070-68232070 on Build GRCh38
rs570270415 Chr.16: 68231992-68231992 on Build GRCh38
rs537285679 Chr.16: 68231993-68231993 on Build GRCh38
rs547507047 Chr.16: 68231872-68231872 on Build GRCh38
rs776765965 Chr.16: 68232014-68232014 on Build GRCh38
rs894939916 Chr.16: 68231915-68231915 on Build GRCh38
rs371881233 Chr.16: 68231905-68231905 on Build GRCh38
rs374327564 Chr.16: 68231869-68231869 on Build GRCh38
rs750972402 Chr.16: 68232050-68232050 on Build GRCh38
rs558333548 Chr.16: 68231991-68231991 on Build GRCh38
rs755162828 Chr.16: 68232071-68232071 on Build GRCh38
rs149234558 Chr.16: 68231987-68231987 on Build GRCh38
rs765108665 Chr.16: 68231947-68231947 on Build GRCh38
rs200064519 Chr.16: 68232060-68232060 on Build GRCh38
rs759468917 Chr.16: 68231934-68231934 on Build GRCh38
rs556204024 Chr.16: 68231994-68231994 on Build GRCh38
rs199522914 Chr.16: 68232027-68232027 on Build GRCh38
rs148578202 Chr.16: 68232012-68232012 on Build GRCh38
rs776296719 Chr.16: 68231932-68231932 on Build GRCh38
rs781429565 Chr.16: 68231868-68231868 on Build GRCh38
rs751362891 Chr.16: 68232015-68232015 on Build GRCh38
rs769751021 Chr.16: 68232016-68232016 on Build GRCh38
rs748194320 Chr.16: 68232011-68232011 on Build GRCh38
rs186017472 Chr.16: 68232049-68232049 on Build GRCh38
rs540156390 Chr.16: 68231966-68231966 on Build GRCh38
rs777968585 Chr.16: 68232085-68232085 on Build GRCh38
rs752446082 Chr.16: 68231948-68231948 on Build GRCh38
rs934245575 Chr.16: 68231933-68231933 on Build GRCh38
rs749493606 Chr.16: 68231882-68231882 on Build GRCh38
rs763741902 Chr.16: 68232028-68232028 on Build GRCh38
rs760552882 Chr.16: 68231913-68231913 on Build GRCh38
rs199619918 Chr.16: 68231935-68231935 on Build GRCh38
rs748303854 Chr.16: 68232078-68232078 on Build GRCh38
rs779263735 Chr.16: 68232074-68232074 on Build GRCh38
rs939637588 Chr.16: 68231874-68231874 on Build GRCh38
rs754144961 Chr.16: 68232068-68232068 on Build GRCh38
rs751027219 Chr.16: 68231974-68231974 on Build GRCh38
rs144444211 Chr.16: 68232009-68232009 on Build GRCh38
rs201908706 Chr.16: 68232043-68232043 on Build GRCh38
rs1010171848 Chr.16: 68232033-68232033 on Build GRCh38
rs12920135 Chr.16: 68231928-68231928 on Build GRCh38
rs369232272 Chr.16: 68231988-68231988 on Build GRCh38
rs761447394 Chr.16: 68232045-68232045 on Build GRCh38
rs763539855 Chr.16: 68231971-68231971 on Build GRCh38

View Full Product Details