Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137437752-137438013 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENTPD8
Entrez Gene ID
377841
Gene Name
ectonucleoside triphosphate diphosphohydrolase 8
Gene Aliases
E-NTPDase, GLSR2492, NTPDase-8, UNQ2492
UniGene
Hs.512562
Gene Chromosome Location
Chr.9: 137434364-137441710 on Build GRCh38
Transcripts
NM_001033113.1 NM_198585.2 XM_011518665.2 XM_011518666.2 XM_011518668.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6497756 c.215T>C p.V72A Chr.9: 137437996-137437996 on Build GRCh38
6497755 c.215T>C p.V72A Chr.9: 137437996-137437996 on Build GRCh38
1107117 c.206C>T p.T69M Chr.9: 137438005-137438005 on Build GRCh38
4565619 c.230_231CC>TT p.A77V Chr.9: 137437980-137437981 on Build GRCh38
3433005 c.198G>T p.E66D Chr.9: 137438013-137438013 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs908870300 Chr.9: 137437802-137437802 on Build GRCh38
rs897013265 Chr.9: 137437834-137437834 on Build GRCh38
rs989106801 Chr.9: 137437915-137437915 on Build GRCh38
rs772402695 Chr.9: 137437794-137437794 on Build GRCh38
rs543970929 Chr.9: 137437853-137437853 on Build GRCh38
rs746181110 Chr.9: 137437952-137437952 on Build GRCh38
rs1013521618 Chr.9: 137437827-137437827 on Build GRCh38
rs537208818 Chr.9: 137437806-137437806 on Build GRCh38
rs200546494 Chr.9: 137437943-137437943 on Build GRCh38
rs200099005 Chr.9: 137437944-137437944 on Build GRCh38
rs748989905 Chr.9: 137437981-137437981 on Build GRCh38
rs758525518 Chr.9: 137437934-137437934 on Build GRCh38
rs963278489 Chr.9: 137437923-137437923 on Build GRCh38
rs1006902581 Chr.9: 137437850-137437850 on Build GRCh38
rs768124732 Chr.9: 137437989-137437989 on Build GRCh38
rs950642154 Chr.9: 137437857-137437857 on Build GRCh38
rs376869050 Chr.9: 137437924-137437924 on Build GRCh38
rs935456022 Chr.9: 137437990-137437990 on Build GRCh38
rs748162594 Chr.9: 137437779-137437779 on Build GRCh38
rs141081294 Chr.9: 137437823-137437823 on Build GRCh38
rs983812793 Chr.9: 137437791-137437791 on Build GRCh38
rs758015896 Chr.9: 137437952-137437952 on Build GRCh38
rs927411941 Chr.9: 137437950-137437950 on Build GRCh38
rs868397070 Chr.9: 137437801-137437801 on Build GRCh38
rs146516878 Chr.9: 137437815-137437815 on Build GRCh38
rs557315236 Chr.9: 137437814-137437814 on Build GRCh38
rs564570975 Chr.9: 137437890-137437901 on Build GRCh38
rs374891120 Chr.9: 137437896-137437896 on Build GRCh38
rs773812361 Chr.9: 137437994-137437994 on Build GRCh38
rs937703858 Chr.9: 137437816-137437816 on Build GRCh38
rs894063085 Chr.9: 137437832-137437832 on Build GRCh38
rs374563315 Chr.9: 137437817-137437817 on Build GRCh38
rs1027108067 Chr.9: 137437847-137437847 on Build GRCh38
rs368312647 Chr.9: 137437955-137437955 on Build GRCh38
rs371503672 Chr.9: 137437925-137437925 on Build GRCh38
rs995736773 Chr.9: 137437842-137437842 on Build GRCh38
rs188781909 Chr.9: 137437831-137437831 on Build GRCh38
rs191126792 Chr.9: 137437895-137437895 on Build GRCh38
rs1018298872 Chr.9: 137437845-137437845 on Build GRCh38
rs909099240 Chr.9: 137437904-137437904 on Build GRCh38
rs954138556 Chr.9: 137437849-137437849 on Build GRCh38
rs200317835 Chr.9: 137437937-137437937 on Build GRCh38
rs753022448 Chr.9: 137437933-137437933 on Build GRCh38
rs937756061 Chr.9: 137437974-137437974 on Build GRCh38
rs778036942 Chr.9: 137437935-137437935 on Build GRCh38
rs779668900 Chr.9: 137437978-137437978 on Build GRCh38
rs183434736 Chr.9: 137437964-137437964 on Build GRCh38

View Full Product Details