Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 119076852-119077109 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VPS11
Entrez Gene ID
55823
Gene Name
VPS11, CORVET/HOPS core subunit
Gene Aliases
END1, HLD12, PEP5, RNF108, hVPS11
UniGene
Hs.234282
Gene Chromosome Location
Chr.11: 119067752-119081978 on Build GRCh38
Transcripts
NM_001290185.1 NM_021729.5 XM_011542898.1 XM_011542899.2

SNP Detail

SNP ID Chromosome Location
rs782757611 Chr.11: 119077033-119077033 on Build GRCh38
rs782478282 Chr.11: 119076952-119076952 on Build GRCh38
rs782136006 Chr.11: 119076902-119076902 on Build GRCh38
rs782792271 Chr.11: 119076923-119076923 on Build GRCh38
rs782751048 Chr.11: 119076917-119076917 on Build GRCh38
rs782429563 Chr.11: 119076931-119076931 on Build GRCh38
rs372388006 Chr.11: 119076985-119076985 on Build GRCh38
rs114313869 Chr.11: 119077061-119077061 on Build GRCh38
rs935117316 Chr.11: 119076893-119076893 on Build GRCh38
rs947731516 Chr.11: 119076966-119076966 on Build GRCh38
rs781868489 Chr.11: 119077048-119077048 on Build GRCh38
rs540261 Chr.11: 119076924-119076924 on Build GRCh38
rs995257266 Chr.11: 119077040-119077040 on Build GRCh38
rs781941159 Chr.11: 119076878-119076878 on Build GRCh38
rs1049107132 Chr.11: 119077041-119077041 on Build GRCh38
rs782250012 Chr.11: 119077011-119077011 on Build GRCh38
rs782157737 Chr.11: 119076885-119076885 on Build GRCh38
rs782073552 Chr.11: 119077050-119077050 on Build GRCh38
rs781848143 Chr.11: 119076918-119076918 on Build GRCh38
rs78868054 Chr.11: 119076881-119076881 on Build GRCh38
rs931213649 Chr.11: 119077013-119077013 on Build GRCh38
rs781958332 Chr.11: 119077023-119077023 on Build GRCh38
rs782462249 Chr.11: 119077062-119077062 on Build GRCh38
rs371834293 Chr.11: 119077029-119077029 on Build GRCh38
rs375711174 Chr.11: 119076889-119076889 on Build GRCh38
rs782410537 Chr.11: 119076999-119076999 on Build GRCh38
rs576059939 Chr.11: 119077026-119077026 on Build GRCh38
rs782696173 Chr.11: 119077027-119077027 on Build GRCh38
rs374855388 Chr.11: 119077059-119077059 on Build GRCh38
rs539555921 Chr.11: 119076932-119076932 on Build GRCh38
rs782393783 Chr.11: 119077012-119077012 on Build GRCh38
rs367865762 Chr.11: 119077007-119077007 on Build GRCh38
rs782253145 Chr.11: 119077076-119077078 on Build GRCh38
rs375848106 Chr.11: 119076995-119076995 on Build GRCh38
rs782442565 Chr.11: 119076920-119076920 on Build GRCh38
rs782507042 Chr.11: 119076926-119076926 on Build GRCh38
rs557547753 Chr.11: 119077017-119077017 on Build GRCh38

View Full Product Details