Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 37413055-37413306 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF569
Entrez Gene ID
148266
Gene Name
zinc finger protein 569
Gene Aliases
ZAP1, ZNF
UniGene
Hs.511848
Gene Chromosome Location
Chr.19: 37411146-37469268 on Build GRCh38
Transcripts
NM_152484.2 XM_006723046.2 XM_006723047.3 XM_006723048.3 XM_011526538.2 XM_011526539.2 XM_017026376.1 XM_017026377.1 XM_017026378.1 XM_017026379.1 XM_017026380.1 XM_017026381.1 XM_017026382.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
304841 c.1412G>T p.C471F Chr.19: 37413246-37413246 on Build GRCh38
6831297 c.1392A>C p.K464N Chr.19: 37413266-37413266 on Build GRCh38
3200839 c.1488C>T p.C496C Chr.19: 37413170-37413170 on Build GRCh38
1630885 c.1387delG p.E463fs*79 Chr.19: 37413271-37413271 on Build GRCh38
3701481 c.1492G>T p.E498* Chr.19: 37413166-37413166 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs184996071 Chr.19: 37413108-37413108 on Build GRCh38
rs752535031 Chr.19: 37413266-37413266 on Build GRCh38
rs751966315 Chr.19: 37413095-37413095 on Build GRCh38
rs367822764 Chr.19: 37413236-37413236 on Build GRCh38
rs767542770 Chr.19: 37413141-37413141 on Build GRCh38
rs750516013 Chr.19: 37413159-37413159 on Build GRCh38
rs181476478 Chr.19: 37413097-37413097 on Build GRCh38
rs756469170 Chr.19: 37413165-37413165 on Build GRCh38
rs760789624 Chr.19: 37413257-37413257 on Build GRCh38
rs777217924 Chr.19: 37413205-37413205 on Build GRCh38
rs778634771 Chr.19: 37413261-37413261 on Build GRCh38
rs867767082 Chr.19: 37413167-37413167 on Build GRCh38
rs776528989 Chr.19: 37413199-37413199 on Build GRCh38
rs746451630 Chr.19: 37413213-37413213 on Build GRCh38
rs140809946 Chr.19: 37413091-37413091 on Build GRCh38
rs763336215 Chr.19: 37413234-37413234 on Build GRCh38
rs780367148 Chr.19: 37413171-37413171 on Build GRCh38
rs772036719 Chr.19: 37413200-37413200 on Build GRCh38
rs768062244 Chr.19: 37413248-37413248 on Build GRCh38
rs1031638418 Chr.19: 37413087-37413087 on Build GRCh38
rs750546547 Chr.19: 37413255-37413255 on Build GRCh38
rs778737687 Chr.19: 37413186-37413186 on Build GRCh38
rs868336826 Chr.19: 37413115-37413115 on Build GRCh38
rs770580566 Chr.19: 37413222-37413222 on Build GRCh38
rs190270449 Chr.19: 37413092-37413092 on Build GRCh38
rs748089245 Chr.19: 37413187-37413187 on Build GRCh38
rs201750077 Chr.19: 37413243-37413243 on Build GRCh38
rs763242908 Chr.19: 37413086-37413086 on Build GRCh38
rs755209171 Chr.19: 37413260-37413260 on Build GRCh38
rs753709721 Chr.19: 37413177-37413177 on Build GRCh38

View Full Product Details