Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 125733343-125733602 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EDRF1
Entrez Gene ID
26098
Gene Name
erythroid differentiation regulatory factor 1
Gene Aliases
C10orf137
UniGene
Hs.468688
Gene Chromosome Location
Chr.10: 125719515-125764143 on Build GRCh38
Transcripts
NM_001202438.1 NM_015608.2 NR_110857.1 NR_110858.1 NR_110859.1 XM_011539622.1 XM_017016066.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6873314 c.1223C>T p.S408L Chr.10: 125733498-125733498 on Build GRCh38
6613837 c.1076A>T p.N359I Chr.10: 125733453-125733453 on Build GRCh38
5630981 c.1201G>A p.D401N Chr.10: 125733476-125733476 on Build GRCh38
6613836 c.1178A>T p.N393I Chr.10: 125733453-125733453 on Build GRCh38
3414793 c.1096A>C p.K366Q Chr.10: 125733473-125733473 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759728204 Chr.10: 125733497-125733497 on Build GRCh38
rs967475911 Chr.10: 125733410-125733410 on Build GRCh38
rs749345289 Chr.10: 125733465-125733467 on Build GRCh38
rs199521946 Chr.10: 125733369-125733369 on Build GRCh38
rs781065557 Chr.10: 125733529-125733529 on Build GRCh38
rs757204140 Chr.10: 125733517-125733517 on Build GRCh38
rs752713079 Chr.10: 125733513-125733513 on Build GRCh38
rs373033431 Chr.10: 125733376-125733376 on Build GRCh38
rs760909693 Chr.10: 125733474-125733474 on Build GRCh38
rs765305161 Chr.10: 125733503-125733503 on Build GRCh38
rs772649125 Chr.10: 125733436-125733436 on Build GRCh38
rs990699861 Chr.10: 125733572-125733572 on Build GRCh38
rs762418966 Chr.10: 125733409-125733409 on Build GRCh38
rs755843851 Chr.10: 125733559-125733559 on Build GRCh38
rs750269268 Chr.10: 125733551-125733551 on Build GRCh38
rs377640681 Chr.10: 125733534-125733534 on Build GRCh38
rs376213703 Chr.10: 125733452-125733452 on Build GRCh38
rs768270288 Chr.10: 125733395-125733395 on Build GRCh38
rs370003289 Chr.10: 125733496-125733496 on Build GRCh38
rs748881280 Chr.10: 125733393-125733393 on Build GRCh38
rs148134506 Chr.10: 125733478-125733478 on Build GRCh38

View Full Product Details