Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 174302036-174302295 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP44
Entrez Gene ID
80817
Gene Name
centrosomal protein 44
Gene Aliases
KIAA1712, PS1TP3
UniGene
Hs.555989
Gene Chromosome Location
Chr.4: 174283677-174334355 on Build GRCh38
Transcripts
NM_001040157.2 NM_001145314.1 XM_011532285.2 XM_011532286.2 XM_011532287.2 XM_017008654.1 XM_017008655.1 XM_017008656.1 XM_017008657.1 XM_017008658.1 XM_017008659.1 XM_017008660.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3602071 c.158A>T p.Y53F Chr.4: 174302107-174302107 on Build GRCh38
5817477 c.191A>G p.E64G Chr.4: 174302140-174302140 on Build GRCh38
6400068 c.200C>A p.A67E Chr.4: 174302149-174302149 on Build GRCh38
1661105 c.185A>T p.N62I Chr.4: 174302134-174302134 on Build GRCh38
4330873 c.208G>T p.D70Y Chr.4: 174302157-174302157 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778278459 Chr.4: 174302108-174302108 on Build GRCh38
rs764500665 Chr.4: 174302216-174302216 on Build GRCh38
rs778418838 Chr.4: 174302234-174302234 on Build GRCh38
rs543619406 Chr.4: 174302095-174302095 on Build GRCh38
rs754423619 Chr.4: 174302222-174302222 on Build GRCh38
rs866493608 Chr.4: 174302070-174302070 on Build GRCh38
rs757713957 Chr.4: 174302230-174302230 on Build GRCh38
rs762939862 Chr.4: 174302156-174302156 on Build GRCh38
rs1045054693 Chr.4: 174302120-174302120 on Build GRCh38
rs372440623 Chr.4: 174302139-174302139 on Build GRCh38
rs201484664 Chr.4: 174302202-174302202 on Build GRCh38
rs906358512 Chr.4: 174302241-174302241 on Build GRCh38
rs368751627 Chr.4: 174302072-174302072 on Build GRCh38
rs755285424 Chr.4: 174302073-174302073 on Build GRCh38
rs775613469 Chr.4: 174302215-174302218 on Build GRCh38
rs759691071 Chr.4: 174302178-174302178 on Build GRCh38
rs568147313 Chr.4: 174302224-174302224 on Build GRCh38
rs776671957 Chr.4: 174302143-174302143 on Build GRCh38
rs143955462 Chr.4: 174302164-174302164 on Build GRCh38
rs907651312 Chr.4: 174302122-174302122 on Build GRCh38
rs1012082066 Chr.4: 174302189-174302189 on Build GRCh38
rs200080536 Chr.4: 174302085-174302085 on Build GRCh38
rs748712778 Chr.4: 174302125-174302125 on Build GRCh38
rs199718601 Chr.4: 174302182-174302182 on Build GRCh38
rs781482160 Chr.4: 174302083-174302083 on Build GRCh38
rs745400483 Chr.4: 174302237-174302237 on Build GRCh38
rs753068909 Chr.4: 174302092-174302092 on Build GRCh38
rs771855113 Chr.4: 174302134-174302134 on Build GRCh38
rs940335470 Chr.4: 174302145-174302145 on Build GRCh38
rs151113549 Chr.4: 174302133-174302133 on Build GRCh38
rs772220005 Chr.4: 174302207-174302209 on Build GRCh38
rs563605142 Chr.4: 174302099-174302099 on Build GRCh38
rs1003406081 Chr.4: 174302253-174302253 on Build GRCh38
rs770538564 Chr.4: 174302130-174302130 on Build GRCh38
rs115721122 Chr.4: 174302163-174302163 on Build GRCh38
rs753013658 Chr.4: 174302184-174302184 on Build GRCh38
rs768690821 Chr.4: 174302142-174302142 on Build GRCh38

View Full Product Details