Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 140632768-140633005 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD14
Entrez Gene ID
929
Gene Name
CD14 molecule
Gene Aliases
-
UniGene
Hs.163867
Gene Chromosome Location
Chr.5: 140631728-140633701 on Build GRCh38
Transcripts
NM_000591.3 NM_001040021.2 NM_001174104.1 NM_001174105.1 XM_011537665.2

Gene Symbol
TMCO6
Entrez Gene ID
55374
Gene Name
transmembrane and coiled-coil domains 6
Gene Aliases
PRO1580
UniGene
Hs.534333
Gene Chromosome Location
Chr.5: 140596529-140647411 on Build GRCh38
Transcripts
NM_000591.3 NM_001040021.2 NM_001174104.1 NM_001174105.1 XM_011537665.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6623399 c.168G>T p.E56D Chr.5: 140632816-140632816 on Build GRCh38
1754019 c.4G>A p.E2K Chr.5: 140632980-140632980 on Build GRCh38
4670338 c.180T>C p.H60H Chr.5: 140632804-140632804 on Build GRCh38
5496714 c.4-10C>T p.? Chr.5: 140632990-140632990 on Build GRCh38
1486340 c.127C>A p.P43T Chr.5: 140632857-140632857 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778777497 Chr.5: 140632800-140632800 on Build GRCh38
rs749780180 Chr.5: 140632940-140632940 on Build GRCh38
rs752685957 Chr.5: 140632946-140632947 on Build GRCh38
rs566040317 Chr.5: 140632922-140632922 on Build GRCh38
rs768967552 Chr.5: 140632834-140632834 on Build GRCh38
rs369079134 Chr.5: 140632799-140632799 on Build GRCh38
rs548438980 Chr.5: 140632936-140632936 on Build GRCh38
rs763569531 Chr.5: 140632860-140632860 on Build GRCh38
rs373361139 Chr.5: 140632875-140632875 on Build GRCh38
rs766163310 Chr.5: 140632956-140632956 on Build GRCh38
rs374046354 Chr.5: 140632805-140632805 on Build GRCh38
rs987397504 Chr.5: 140632980-140632980 on Build GRCh38
rs766166161 Chr.5: 140632788-140632788 on Build GRCh38
rs530033380 Chr.5: 140632930-140632930 on Build GRCh38
rs774742461 Chr.5: 140632951-140632951 on Build GRCh38
rs780169248 Chr.5: 140632821-140632821 on Build GRCh38
rs760222076 Chr.5: 140632867-140632867 on Build GRCh38
rs765027621 Chr.5: 140632947-140632949 on Build GRCh38
rs758551783 Chr.5: 140632959-140632960 on Build GRCh38
rs761598550 Chr.5: 140632881-140632881 on Build GRCh38
rs765014968 Chr.5: 140632977-140632977 on Build GRCh38
rs781436427 Chr.5: 140632925-140632925 on Build GRCh38
rs151227107 Chr.5: 140632794-140632794 on Build GRCh38
rs758412333 Chr.5: 140632806-140632806 on Build GRCh38
rs748324474 Chr.5: 140632928-140632928 on Build GRCh38
rs770314128 Chr.5: 140632852-140632852 on Build GRCh38
rs746483705 Chr.5: 140632958-140632958 on Build GRCh38
rs768052444 Chr.5: 140632963-140632963 on Build GRCh38
rs569831840 Chr.5: 140632945-140632945 on Build GRCh38
rs147037491 Chr.5: 140632854-140632854 on Build GRCh38
rs3138079 Chr.5: 140632969-140632969 on Build GRCh38
rs753538025 Chr.5: 140632802-140632810 on Build GRCh38
rs776285862 Chr.5: 140632971-140632971 on Build GRCh38
rs375798957 Chr.5: 140632904-140632904 on Build GRCh38
rs761427509 Chr.5: 140632972-140632972 on Build GRCh38
rs141411678 Chr.5: 140632830-140632830 on Build GRCh38
rs368600149 Chr.5: 140632912-140632912 on Build GRCh38
rs775178447 Chr.5: 140632866-140632866 on Build GRCh38
rs765027427 Chr.5: 140632887-140632887 on Build GRCh38
rs906360518 Chr.5: 140632954-140632954 on Build GRCh38
rs547856424 Chr.5: 140632841-140632841 on Build GRCh38
rs1047799644 Chr.5: 140632812-140632812 on Build GRCh38
rs780081235 Chr.5: 140632903-140632903 on Build GRCh38
rs750362237 Chr.5: 140632803-140632803 on Build GRCh38
rs138255654 Chr.5: 140632861-140632861 on Build GRCh38
rs759399709 Chr.5: 140632926-140632927 on Build GRCh38
rs543735754 Chr.5: 140632862-140632862 on Build GRCh38
rs758399990 Chr.5: 140632899-140632899 on Build GRCh38
rs750343901 Chr.5: 140632893-140632893 on Build GRCh38
rs911909690 Chr.5: 140632981-140632981 on Build GRCh38
rs763753720 Chr.5: 140632873-140632873 on Build GRCh38

View Full Product Details