Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 57130466-57130711 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USP29
Entrez Gene ID
57663
Gene Name
ubiquitin specific peptidase 29
Gene Aliases
HOM-TES-84/86
UniGene
Hs.515632
Gene Chromosome Location
Chr.19: 57120141-57131926 on Build GRCh38
Transcripts
NM_020903.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1002083 c.1864C>A p.L622M Chr.19: 57130539-57130539 on Build GRCh38
6570732 c.1924G>A p.E642K Chr.19: 57130599-57130599 on Build GRCh38
1002084 c.1897G>A p.V633I Chr.19: 57130572-57130572 on Build GRCh38
5856890 c.1996G>A p.G666R Chr.19: 57130671-57130671 on Build GRCh38
4533705 c.2007G>A p.L669L Chr.19: 57130682-57130682 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs543467918 Chr.19: 57130596-57130596 on Build GRCh38
rs868065210 Chr.19: 57130599-57130599 on Build GRCh38
rs200311952 Chr.19: 57130629-57130629 on Build GRCh38
rs553671474 Chr.19: 57130500-57130500 on Build GRCh38
rs777494963 Chr.19: 57130645-57130645 on Build GRCh38
rs770876996 Chr.19: 57130564-57130564 on Build GRCh38
rs375010576 Chr.19: 57130593-57130593 on Build GRCh38
rs758853885 Chr.19: 57130505-57130505 on Build GRCh38
rs772991405 Chr.19: 57130527-57130528 on Build GRCh38
rs750445433 Chr.19: 57130597-57130597 on Build GRCh38
rs774875326 Chr.19: 57130671-57130671 on Build GRCh38
rs771973122 Chr.19: 57130552-57130552 on Build GRCh38
rs576803775 Chr.19: 57130559-57130559 on Build GRCh38
rs764495368 Chr.19: 57130592-57130592 on Build GRCh38
rs541335351 Chr.19: 57130549-57130549 on Build GRCh38
rs373629916 Chr.19: 57130513-57130513 on Build GRCh38
rs770825632 Chr.19: 57130660-57130660 on Build GRCh38
rs762366118 Chr.19: 57130595-57130595 on Build GRCh38
rs773000243 Chr.19: 57130557-57130557 on Build GRCh38
rs1011633218 Chr.19: 57130635-57130635 on Build GRCh38
rs756255665 Chr.19: 57130526-57130526 on Build GRCh38
rs563272011 Chr.19: 57130609-57130609 on Build GRCh38
rs764776153 Chr.19: 57130588-57130588 on Build GRCh38
rs754028995 Chr.19: 57130622-57130622 on Build GRCh38
rs918604342 Chr.19: 57130514-57130514 on Build GRCh38
rs866020470 Chr.19: 57130542-57130542 on Build GRCh38
rs780137813 Chr.19: 57130616-57130616 on Build GRCh38
rs754319159 Chr.19: 57130620-57130620 on Build GRCh38
rs769280240 Chr.19: 57130669-57130669 on Build GRCh38
rs1037391516 Chr.19: 57130525-57130525 on Build GRCh38
rs781368378 Chr.19: 57130521-57130521 on Build GRCh38
rs911280997 Chr.19: 57130551-57130551 on Build GRCh38
rs1041260054 Chr.19: 57130586-57130586 on Build GRCh38
rs780370092 Chr.19: 57130529-57130529 on Build GRCh38
rs35436572 Chr.19: 57130591-57130591 on Build GRCh38
rs764691850 Chr.19: 57130507-57130507 on Build GRCh38
rs745363541 Chr.19: 57130665-57130665 on Build GRCh38
rs746837976 Chr.19: 57130651-57130651 on Build GRCh38
rs148895356 Chr.19: 57130688-57130688 on Build GRCh38
rs776080625 Chr.19: 57130566-57130566 on Build GRCh38
rs764973830 Chr.19: 57130490-57130490 on Build GRCh38
rs911918524 Chr.19: 57130501-57130501 on Build GRCh38
rs759127453 Chr.19: 57130578-57130578 on Build GRCh38
rs765404620 Chr.19: 57130503-57130504 on Build GRCh38
rs1035334665 Chr.19: 57130598-57130598 on Build GRCh38
rs781241321 Chr.19: 57130661-57130661 on Build GRCh38
rs74511605 Chr.19: 57130519-57130519 on Build GRCh38

View Full Product Details