Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 81702644-81702875 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZFAND1
Entrez Gene ID
79752
Gene Name
zinc finger AN1-type containing 1
Gene Aliases
-
UniGene
Hs.655453
Gene Chromosome Location
Chr.8: 81701331-81721304 on Build GRCh38
Transcripts
NM_001170796.1 NM_024699.2 NR_033193.1 NR_033194.1 NR_033195.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4917063 c.683A>G p.D228G Chr.8: 81702819-81702819 on Build GRCh38
7126370 c.715G>C p.D239H Chr.8: 81702787-81702787 on Build GRCh38
6114251 c.709A>T p.K237* Chr.8: 81702793-81702793 on Build GRCh38
3942788 c.741T>G p.N247K Chr.8: 81702761-81702761 on Build GRCh38
1289038 c.787G>A p.V263I Chr.8: 81702715-81702715 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201469575 Chr.8: 81702691-81702691 on Build GRCh38
rs930731089 Chr.8: 81702829-81702829 on Build GRCh38
rs149747946 Chr.8: 81702775-81702775 on Build GRCh38
rs781275964 Chr.8: 81702837-81702837 on Build GRCh38
rs377085653 Chr.8: 81702843-81702843 on Build GRCh38
rs758742596 Chr.8: 81702796-81702796 on Build GRCh38
rs1028787559 Chr.8: 81702733-81702733 on Build GRCh38
rs758980819 Chr.8: 81702743-81702744 on Build GRCh38
rs762812128 Chr.8: 81702693-81702693 on Build GRCh38
rs763681832 Chr.8: 81702779-81702779 on Build GRCh38
rs755445180 Chr.8: 81702824-81702824 on Build GRCh38
rs747512116 Chr.8: 81702814-81702814 on Build GRCh38
rs766327906 Chr.8: 81702704-81702704 on Build GRCh38
rs776588533 Chr.8: 81702826-81702826 on Build GRCh38
rs769955968 Chr.8: 81702840-81702840 on Build GRCh38
rs774419977 Chr.8: 81702717-81702717 on Build GRCh38
rs764819894 Chr.8: 81702851-81702852 on Build GRCh38
rs780456491 Chr.8: 81702812-81702812 on Build GRCh38
rs928814242 Chr.8: 81702692-81702692 on Build GRCh38
rs576125001 Chr.8: 81702820-81702820 on Build GRCh38
rs527774502 Chr.8: 81702790-81702790 on Build GRCh38
rs546228778 Chr.8: 81702839-81702839 on Build GRCh38
rs752166418 Chr.8: 81702741-81702741 on Build GRCh38
rs759617687 Chr.8: 81702728-81702728 on Build GRCh38
rs766988787 Chr.8: 81702734-81702734 on Build GRCh38
rs776140486 Chr.8: 81702717-81702717 on Build GRCh38
rs1036798701 Chr.8: 81702682-81702682 on Build GRCh38

View Full Product Details