Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 123630126-123630371 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF2B1
Entrez Gene ID
1967
Gene Name
eukaryotic translation initiation factor 2B subunit alpha
Gene Aliases
EIF2B, EIF2BA
UniGene
Hs.741273
Gene Chromosome Location
Chr.12: 123621023-123633776 on Build GRCh38
Transcripts
NM_001414.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1605720 c.265T>C p.C89R Chr.12: 123630273-123630273 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs944357736 Chr.12: 123630151-123630151 on Build GRCh38
rs746822849 Chr.12: 123630308-123630308 on Build GRCh38
rs757985338 Chr.12: 123630246-123630246 on Build GRCh38
rs374302327 Chr.12: 123630307-123630307 on Build GRCh38
rs747091269 Chr.12: 123630150-123630152 on Build GRCh38
rs863225050 Chr.12: 123630210-123630210 on Build GRCh38
rs142755878 Chr.12: 123630257-123630257 on Build GRCh38
rs770912605 Chr.12: 123630173-123630173 on Build GRCh38
rs759728057 Chr.12: 123630193-123630193 on Build GRCh38
rs781471132 Chr.12: 123630253-123630254 on Build GRCh38
rs765287085 Chr.12: 123630205-123630205 on Build GRCh38
rs1007863823 Chr.12: 123630158-123630158 on Build GRCh38
rs1050397 Chr.12: 123630252-123630252 on Build GRCh38
rs747617594 Chr.12: 123630265-123630265 on Build GRCh38
rs775739205 Chr.12: 123630217-123630217 on Build GRCh38
rs553282463 Chr.12: 123630154-123630154 on Build GRCh38
rs1041124768 Chr.12: 123630170-123630170 on Build GRCh38
rs148513104 Chr.12: 123630167-123630167 on Build GRCh38
rs757509025 Chr.12: 123630248-123630248 on Build GRCh38
rs755054824 Chr.12: 123630157-123630157 on Build GRCh38
rs746730077 Chr.12: 123630169-123630169 on Build GRCh38
rs768076568 Chr.12: 123630249-123630249 on Build GRCh38
rs776365178 Chr.12: 123630190-123630190 on Build GRCh38
rs773170410 Chr.12: 123630165-123630165 on Build GRCh38
rs778778741 Chr.12: 123630146-123630146 on Build GRCh38
rs748375449 Chr.12: 123630255-123630256 on Build GRCh38
rs770625902 Chr.12: 123630312-123630312 on Build GRCh38
rs781247492 Chr.12: 123630317-123630317 on Build GRCh38
rs764262014 Chr.12: 123630218-123630218 on Build GRCh38
rs758142806 Chr.12: 123630269-123630269 on Build GRCh38
rs930417910 Chr.12: 123630332-123630332 on Build GRCh38

View Full Product Details