Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 53871145-53871406 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTR8
Entrez Gene ID
93973
Gene Name
ARP8 actin-related protein 8 homolog
Gene Aliases
ARP8, INO80N, hArp8
UniGene
Hs.412186
Gene Chromosome Location
Chr.3: 53863096-53882202 on Build GRCh38
Transcripts
NM_022899.4 XM_005265587.4 XM_011534249.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7415388 c.566G>A p.G189D Chr.3: 53871348-53871348 on Build GRCh38
1309382 c.558G>T p.L186L Chr.3: 53871356-53871356 on Build GRCh38
1660385 c.525G>T p.E175D Chr.3: 53871389-53871389 on Build GRCh38
1660383 c.674A>G p.D225G Chr.3: 53871240-53871240 on Build GRCh38
6836067 c.1498G>A p.A500T Chr.3: 53871301-53871301 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751785539 Chr.3: 53871225-53871225 on Build GRCh38
rs199775265 Chr.3: 53871366-53871366 on Build GRCh38
rs373055423 Chr.3: 53871316-53871316 on Build GRCh38
rs759505789 Chr.3: 53871192-53871192 on Build GRCh38
rs757458013 Chr.3: 53871239-53871239 on Build GRCh38
rs375589353 Chr.3: 53871376-53871376 on Build GRCh38
rs371208984 Chr.3: 53871266-53871266 on Build GRCh38
rs199774315 Chr.3: 53871211-53871211 on Build GRCh38
rs868041537 Chr.3: 53871280-53871281 on Build GRCh38
rs765208723 Chr.3: 53871193-53871193 on Build GRCh38
rs922515284 Chr.3: 53871287-53871287 on Build GRCh38
rs569041085 Chr.3: 53871332-53871332 on Build GRCh38
rs751374614 Chr.3: 53871331-53871331 on Build GRCh38
rs535696904 Chr.3: 53871270-53871270 on Build GRCh38
rs957771811 Chr.3: 53871240-53871240 on Build GRCh38
rs1046677 Chr.3: 53871281-53871281 on Build GRCh38
rs969154827 Chr.3: 53871272-53871272 on Build GRCh38
rs778651695 Chr.3: 53871183-53871185 on Build GRCh38
rs774354335 Chr.3: 53871381-53871381 on Build GRCh38
rs769465138 Chr.3: 53871300-53871300 on Build GRCh38
rs758248037 Chr.3: 53871251-53871251 on Build GRCh38
rs376181693 Chr.3: 53871343-53871343 on Build GRCh38
rs768958871 Chr.3: 53871269-53871269 on Build GRCh38
rs750522691 Chr.3: 53871249-53871249 on Build GRCh38
rs1012882579 Chr.3: 53871197-53871197 on Build GRCh38
rs775522509 Chr.3: 53871304-53871304 on Build GRCh38
rs749863203 Chr.3: 53871224-53871224 on Build GRCh38
rs767573812 Chr.3: 53871338-53871338 on Build GRCh38
rs750658174 Chr.3: 53871340-53871340 on Build GRCh38
rs758904462 Chr.3: 53871298-53871298 on Build GRCh38
rs762976722 Chr.3: 53871371-53871371 on Build GRCh38
rs770548322 Chr.3: 53871293-53871293 on Build GRCh38
rs779124592 Chr.3: 53871210-53871210 on Build GRCh38
rs556990155 Chr.3: 53871344-53871344 on Build GRCh38
rs776360774 Chr.3: 53871294-53871294 on Build GRCh38
rs796521446 Chr.3: 53871250-53871250 on Build GRCh38
rs368263409 Chr.3: 53871365-53871365 on Build GRCh38
rs748405850 Chr.3: 53871353-53871353 on Build GRCh38
rs999146302 Chr.3: 53871222-53871222 on Build GRCh38
rs780472666 Chr.3: 53871264-53871264 on Build GRCh38
rs779325108 Chr.3: 53871350-53871350 on Build GRCh38
rs752689478 Chr.3: 53871209-53871209 on Build GRCh38
rs781324307 Chr.3: 53871248-53871248 on Build GRCh38
rs769409988 Chr.3: 53871370-53871370 on Build GRCh38
rs201085829 Chr.3: 53871356-53871356 on Build GRCh38
rs533271957 Chr.3: 53871349-53871349 on Build GRCh38
rs1030661035 Chr.3: 53871230-53871230 on Build GRCh38
rs980673721 Chr.3: 53871275-53871275 on Build GRCh38
rs763953547 Chr.3: 53871320-53871320 on Build GRCh38

View Full Product Details