Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 74755812-74756071 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTC
Entrez Gene ID
685
Gene Name
betacellulin
Gene Aliases
-
UniGene
Hs.591704
Gene Chromosome Location
Chr.4: 74744759-74794718 on Build GRCh38
Transcripts
NM_001316963.1 NM_001729.3 XM_011532211.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2831509 c.269C>T p.T90M Chr.4: 74755871-74755871 on Build GRCh38
4398186 c.240G>A p.G80G Chr.4: 74755900-74755900 on Build GRCh38
4389585 c.164-9A>G p.? Chr.4: 74755985-74755985 on Build GRCh38
343678 c.192C>T p.G64G Chr.4: 74755948-74755948 on Build GRCh38
5793332 c.281+8G>C p.? Chr.4: 74755851-74755851 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201123135 Chr.4: 74755958-74755958 on Build GRCh38
rs782656231 Chr.4: 74755869-74755869 on Build GRCh38
rs141442985 Chr.4: 74755871-74755871 on Build GRCh38
rs375367986 Chr.4: 74755955-74755955 on Build GRCh38
rs551672350 Chr.4: 74755879-74755879 on Build GRCh38
rs782328196 Chr.4: 74755987-74755987 on Build GRCh38
rs782165620 Chr.4: 74755935-74755935 on Build GRCh38
rs782000718 Chr.4: 74756021-74756021 on Build GRCh38
rs943311509 Chr.4: 74755847-74755847 on Build GRCh38
rs781860037 Chr.4: 74755856-74755856 on Build GRCh38
rs574624337 Chr.4: 74755870-74755870 on Build GRCh38
rs534728400 Chr.4: 74755954-74755954 on Build GRCh38
rs146673863 Chr.4: 74755925-74755925 on Build GRCh38
rs781925130 Chr.4: 74755902-74755902 on Build GRCh38
rs150258991 Chr.4: 74755910-74755910 on Build GRCh38
rs782704683 Chr.4: 74755945-74755945 on Build GRCh38
rs782615265 Chr.4: 74755996-74755996 on Build GRCh38
rs782749174 Chr.4: 74755843-74755843 on Build GRCh38
rs782691067 Chr.4: 74755957-74755957 on Build GRCh38
rs772843382 Chr.4: 74755857-74755857 on Build GRCh38
rs782514658 Chr.4: 74755956-74755956 on Build GRCh38
rs574046461 Chr.4: 74755952-74755952 on Build GRCh38
rs782589887 Chr.4: 74755878-74755878 on Build GRCh38
rs560139741 Chr.4: 74755907-74755907 on Build GRCh38
rs538759116 Chr.4: 74755839-74755839 on Build GRCh38
rs782289206 Chr.4: 74755888-74755888 on Build GRCh38
rs782353910 Chr.4: 74755881-74755881 on Build GRCh38
rs781804250 Chr.4: 74755947-74755947 on Build GRCh38
rs781972691 Chr.4: 74755912-74755912 on Build GRCh38
rs1011841835 Chr.4: 74755941-74755941 on Build GRCh38
rs372117425 Chr.4: 74755893-74755893 on Build GRCh38
rs782781797 Chr.4: 74755922-74755922 on Build GRCh38
rs781810547 Chr.4: 74755834-74755834 on Build GRCh38
rs1039155018 Chr.4: 74755862-74755862 on Build GRCh38
rs781919821 Chr.4: 74756026-74756026 on Build GRCh38
rs782518586 Chr.4: 74755858-74755858 on Build GRCh38
rs911888449 Chr.4: 74755837-74755837 on Build GRCh38
rs782191240 Chr.4: 74755960-74755960 on Build GRCh38
rs782064923 Chr.4: 74756028-74756028 on Build GRCh38
rs987124092 Chr.4: 74756044-74756044 on Build GRCh38
rs576808222 Chr.4: 74755948-74755948 on Build GRCh38
rs782417348 Chr.4: 74756025-74756025 on Build GRCh38
rs11734944 Chr.4: 74756007-74756007 on Build GRCh38
rs201624037 Chr.4: 74756022-74756022 on Build GRCh38
rs199674256 Chr.4: 74755892-74755892 on Build GRCh38

View Full Product Details