Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144423656-144423893 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VPS28
Entrez Gene ID
51160
Gene Name
VPS28, ESCRT-I subunit
Gene Aliases
-
UniGene
Hs.418175
Gene Chromosome Location
Chr.8: 144423601-144428563 on Build GRCh38
Transcripts
NM_016208.3 NM_183057.2 XM_005272323.3 XM_005272324.3 XM_005272325.3 XM_011517099.1 XM_011517100.1 XM_017013529.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7180484 c.605G>C p.R202P Chr.8: 144423866-144423866 on Build GRCh38
6516849 c.640G>T p.A214S Chr.8: 144423831-144423831 on Build GRCh38
5467372 c.654C>T p.F218F Chr.8: 144423817-144423817 on Build GRCh38
6536699 c.609G>C p.Q203H Chr.8: 144423862-144423862 on Build GRCh38
4503949 c.651C>T p.R217R Chr.8: 144423820-144423820 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782274158 Chr.8: 144423733-144423733 on Build GRCh38
rs781809209 Chr.8: 144423824-144423824 on Build GRCh38
rs534218543 Chr.8: 144423702-144423702 on Build GRCh38
rs782028836 Chr.8: 144423793-144423793 on Build GRCh38
rs782362007 Chr.8: 144423788-144423788 on Build GRCh38
rs782522312 Chr.8: 144423756-144423756 on Build GRCh38
rs782633289 Chr.8: 144423841-144423841 on Build GRCh38
rs782556531 Chr.8: 144423820-144423820 on Build GRCh38
rs368968346 Chr.8: 144423768-144423768 on Build GRCh38
rs376242912 Chr.8: 144423801-144423801 on Build GRCh38
rs939256309 Chr.8: 144423850-144423850 on Build GRCh38
rs782367676 Chr.8: 144423774-144423774 on Build GRCh38
rs752498443 Chr.8: 144423712-144423712 on Build GRCh38
rs10527 Chr.8: 144423802-144423802 on Build GRCh38
rs11948 Chr.8: 144423755-144423755 on Build GRCh38
rs782726132 Chr.8: 144423796-144423796 on Build GRCh38
rs556590841 Chr.8: 144423747-144423747 on Build GRCh38
rs545259517 Chr.8: 144423791-144423791 on Build GRCh38
rs760551691 Chr.8: 144423803-144423803 on Build GRCh38
rs1037899835 Chr.8: 144423676-144423676 on Build GRCh38
rs782293340 Chr.8: 144423867-144423867 on Build GRCh38
rs899017121 Chr.8: 144423684-144423684 on Build GRCh38
rs368501757 Chr.8: 144423832-144423832 on Build GRCh38
rs782641635 Chr.8: 144423760-144423760 on Build GRCh38
rs371271081 Chr.8: 144423783-144423783 on Build GRCh38
rs1033377474 Chr.8: 144423722-144423722 on Build GRCh38
rs375225604 Chr.8: 144423822-144423822 on Build GRCh38
rs781787964 Chr.8: 144423797-144423797 on Build GRCh38
rs1003209561 Chr.8: 144423714-144423714 on Build GRCh38
rs140103128 Chr.8: 144423853-144423853 on Build GRCh38
rs374698280 Chr.8: 144423790-144423790 on Build GRCh38
rs1047667985 Chr.8: 144423703-144423703 on Build GRCh38
rs894851641 Chr.8: 144423732-144423732 on Build GRCh38
rs116517218 Chr.8: 144423762-144423762 on Build GRCh38
rs567607459 Chr.8: 144423728-144423728 on Build GRCh38
rs1010655511 Chr.8: 144423739-144423739 on Build GRCh38
rs782012319 Chr.8: 144423791-144423791 on Build GRCh38
rs968926432 Chr.8: 144423745-144423745 on Build GRCh38
rs781963698 Chr.8: 144423781-144423781 on Build GRCh38
rs11558161 Chr.8: 144423794-144423794 on Build GRCh38
rs782625320 Chr.8: 144423856-144423856 on Build GRCh38
rs782294422 Chr.8: 144423761-144423761 on Build GRCh38
rs781860283 Chr.8: 144423812-144423812 on Build GRCh38
rs555342032 Chr.8: 144423705-144423705 on Build GRCh38
rs537967115 Chr.8: 144423740-144423740 on Build GRCh38

View Full Product Details