Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48211589-48211862 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CARD8
Entrez Gene ID
22900
Gene Name
caspase recruitment domain family member 8
Gene Aliases
CARDINAL, DACAR, DAKAR, NDPP, NDPP1, TUCAN
UniGene
Hs.446146
Gene Chromosome Location
Chr.19: 48203148-48256269 on Build GRCh38
Transcripts
NM_001184900.1 NM_001184901.1 NM_001184902.1 NM_001184903.1 NM_014959.3 NR_033678.1 NR_033679.1 NR_033680.1 XM_006723091.3 XM_006723092.3 XM_006723093.3 XM_006723094.3 XM_006723095.3 XM_006723096.3 XM_006723097.3 XM_006723098.3 XM_006723099.3 XM_006723101.2 XM_006723102.3 XM_006723104.3 XM_006723106.3 XM_006723107.3 XM_006723109.3 XM_011526641.2 XM_011526642.2 XM_011526643.2 XM_011526644.2 XM_011526646.2 XM_011526647.2 XM_011526648.2 XM_011526650.2 XM_011526653.2 XM_017026482.1 XM_017026483.1 XM_017026484.1 XM_017026485.1 XM_017026486.1 XM_017026487.1 XM_017026488.1 XM_017026489.1 XM_017026490.1 XM_017026492.1 XM_017026493.1 XM_017026494.1 XM_017026495.1 XM_017026496.1 XM_017026497.1 XM_017026498.1 XM_017026499.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7129946 c.1276C>T p.L426F Chr.19: 48211730-48211730 on Build GRCh38
7201010 c.1593T>C p.Y531Y Chr.19: 48211731-48211731 on Build GRCh38
1712545 c.1601A>C p.Q534P Chr.19: 48211723-48211723 on Build GRCh38
7201056 c.1596T>C p.L532L Chr.19: 48211728-48211728 on Build GRCh38
1729866 c.1496A>G p.N499S Chr.19: 48211828-48211828 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773435031 Chr.19: 48211699-48211699 on Build GRCh38
rs149162452 Chr.19: 48211840-48211840 on Build GRCh38
rs749382283 Chr.19: 48211700-48211700 on Build GRCh38
rs765678614 Chr.19: 48211737-48211737 on Build GRCh38
rs775792481 Chr.19: 48211733-48211733 on Build GRCh38
rs760025108 Chr.19: 48211696-48211699 on Build GRCh38
rs771805440 Chr.19: 48211677-48211677 on Build GRCh38
rs756786481 Chr.19: 48211751-48211751 on Build GRCh38
rs754800916 Chr.19: 48211778-48211778 on Build GRCh38
rs767889684 Chr.19: 48211719-48211719 on Build GRCh38
rs140428405 Chr.19: 48211740-48211740 on Build GRCh38
rs768643736 Chr.19: 48211688-48211688 on Build GRCh38
rs756792561 Chr.19: 48211757-48211757 on Build GRCh38
rs767068052 Chr.19: 48211765-48211766 on Build GRCh38
rs761074966 Chr.19: 48211822-48211822 on Build GRCh38
rs755929405 Chr.19: 48211678-48211678 on Build GRCh38
rs758006827 Chr.19: 48211766-48211766 on Build GRCh38
rs368359327 Chr.19: 48211803-48211803 on Build GRCh38
rs8112588 Chr.19: 48211779-48211779 on Build GRCh38
rs746082189 Chr.19: 48211801-48211801 on Build GRCh38
rs371660482 Chr.19: 48211667-48211667 on Build GRCh38
rs35920934 Chr.19: 48211775-48211775 on Build GRCh38
rs779645807 Chr.19: 48211633-48211633 on Build GRCh38
rs764776725 Chr.19: 48211752-48211752 on Build GRCh38
rs746969682 Chr.19: 48211686-48211686 on Build GRCh38
rs756075950 Chr.19: 48211841-48211842 on Build GRCh38
rs749260047 Chr.19: 48211784-48211784 on Build GRCh38
rs558188524 Chr.19: 48211814-48211814 on Build GRCh38
rs550969694 Chr.19: 48211664-48211664 on Build GRCh38
rs190997806 Chr.19: 48211749-48211749 on Build GRCh38
rs182337605 Chr.19: 48211701-48211701 on Build GRCh38
rs764409113 Chr.19: 48211831-48211831 on Build GRCh38
rs767222799 Chr.19: 48211630-48211630 on Build GRCh38
rs995434554 Chr.19: 48211811-48211811 on Build GRCh38
rs749851325 Chr.19: 48211796-48211797 on Build GRCh38
rs564491556 Chr.19: 48211668-48211668 on Build GRCh38
rs898139743 Chr.19: 48211808-48211808 on Build GRCh38
rs999862762 Chr.19: 48211837-48211837 on Build GRCh38
rs774431953 Chr.19: 48211787-48211787 on Build GRCh38
rs549570428 Chr.19: 48211768-48211768 on Build GRCh38
rs146880951 Chr.19: 48211739-48211739 on Build GRCh38
rs759781205 Chr.19: 48211713-48211713 on Build GRCh38
rs770975518 Chr.19: 48211785-48211785 on Build GRCh38
rs375359387 Chr.19: 48211672-48211672 on Build GRCh38
rs908522427 Chr.19: 48211637-48211637 on Build GRCh38
rs751288332 Chr.19: 48211743-48211744 on Build GRCh38
rs750097954 Chr.19: 48211755-48211755 on Build GRCh38
rs533501140 Chr.19: 48211671-48211671 on Build GRCh38
rs146657505 Chr.19: 48211841-48211841 on Build GRCh38
rs774474455 Chr.19: 48211704-48211704 on Build GRCh38

View Full Product Details