Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32775876-32776137 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YARS
Entrez Gene ID
8565
Gene Name
tyrosyl-tRNA synthetase
Gene Aliases
CMTDIC, TYRRS, YRS, YTS
UniGene
Hs.213264
Gene Chromosome Location
Chr.1: 32775237-32818032 on Build GRCh38
Transcripts
NM_003680.3 XM_011542347.1 XM_011542348.1 XM_017002651.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
908344 c.1526A>G p.N509S Chr.1: 32776042-32776042 on Build GRCh38
4031524 c.1478C>T p.A493V Chr.1: 32776090-32776090 on Build GRCh38
1341854 c.1575delG p.N526fs*>3 Chr.1: 32775993-32775993 on Build GRCh38
4893792 c.1543G>A p.G515S Chr.1: 32776025-32776025 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs139496156 Chr.1: 32776005-32776005 on Build GRCh38
rs752850545 Chr.1: 32776043-32776043 on Build GRCh38
rs774884649 Chr.1: 32775984-32775984 on Build GRCh38
rs769566379 Chr.1: 32776098-32776098 on Build GRCh38
rs777955605 Chr.1: 32775963-32775963 on Build GRCh38
rs780648145 Chr.1: 32776108-32776109 on Build GRCh38
rs781560790 Chr.1: 32775961-32775961 on Build GRCh38
rs749596742 Chr.1: 32776062-32776062 on Build GRCh38
rs756253717 Chr.1: 32776045-32776045 on Build GRCh38
rs746302434 Chr.1: 32776067-32776067 on Build GRCh38
rs761511054 Chr.1: 32776097-32776097 on Build GRCh38
rs201687117 Chr.1: 32775997-32775997 on Build GRCh38
rs371891919 Chr.1: 32776109-32776109 on Build GRCh38
rs943186269 Chr.1: 32775993-32775993 on Build GRCh38
rs924396529 Chr.1: 32775904-32775904 on Build GRCh38
rs757695852 Chr.1: 32776064-32776064 on Build GRCh38
rs200375230 Chr.1: 32776082-32776082 on Build GRCh38
rs771314633 Chr.1: 32775977-32775977 on Build GRCh38
rs768160751 Chr.1: 32776081-32776081 on Build GRCh38
rs933078773 Chr.1: 32775912-32775912 on Build GRCh38
rs749697240 Chr.1: 32775965-32775965 on Build GRCh38
rs141974881 Chr.1: 32775998-32775998 on Build GRCh38
rs772578261 Chr.1: 32776068-32776068 on Build GRCh38
rs771633110 Chr.1: 32775991-32775991 on Build GRCh38
rs774931513 Chr.1: 32776072-32776072 on Build GRCh38
rs371220818 Chr.1: 32776019-32776019 on Build GRCh38
rs887070267 Chr.1: 32776058-32776058 on Build GRCh38
rs750454629 Chr.1: 32776004-32776004 on Build GRCh38
rs746559511 Chr.1: 32776074-32776074 on Build GRCh38
rs758807329 Chr.1: 32775960-32775960 on Build GRCh38
rs762938417 Chr.1: 32776105-32776105 on Build GRCh38
rs774982567 Chr.1: 32775994-32775994 on Build GRCh38
rs879254282 Chr.1: 32776025-32776025 on Build GRCh38
rs750871661 Chr.1: 32775949-32775959 on Build GRCh38
rs765073056 Chr.1: 32776001-32776001 on Build GRCh38
rs1022517973 Chr.1: 32775900-32775900 on Build GRCh38
rs773154497 Chr.1: 32776102-32776102 on Build GRCh38
rs149812692 Chr.1: 32775960-32775960 on Build GRCh38
rs367622240 Chr.1: 32776088-32776088 on Build GRCh38
rs751669673 Chr.1: 32775947-32775947 on Build GRCh38
rs765250908 Chr.1: 32775934-32775934 on Build GRCh38
rs745367045 Chr.1: 32775989-32775989 on Build GRCh38
rs145242872 Chr.1: 32776065-32776065 on Build GRCh38
rs1035749824 Chr.1: 32776066-32776066 on Build GRCh38
rs778099655 Chr.1: 32776061-32776061 on Build GRCh38

View Full Product Details