Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 162374380-162374643 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf111
Entrez Gene ID
284680
Gene Name
chromosome 1 open reading frame 111
Gene Aliases
HSD20
UniGene
Hs.97784
Gene Chromosome Location
Chr.1: 162373725-162376962 on Build GRCh38
Transcripts
NM_182581.3 XM_005245103.3 XM_011509443.2 XM_011509444.1 XM_011509445.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4537760 c.245G>A p.G82E Chr.1: 162374589-162374589 on Build GRCh38
4140905 c.337G>A p.A113T Chr.1: 162374497-162374497 on Build GRCh38
676595 c.335A>C p.E112A Chr.1: 162374499-162374499 on Build GRCh38
4024504 c.307G>A p.V103M Chr.1: 162374527-162374527 on Build GRCh38
6615042 c.237C>T p.T79T Chr.1: 162374597-162374597 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs541661739 Chr.1: 162374477-162374477 on Build GRCh38
rs368265905 Chr.1: 162374470-162374470 on Build GRCh38
rs375087871 Chr.1: 162374598-162374598 on Build GRCh38
rs763508901 Chr.1: 162374614-162374614 on Build GRCh38
rs745670990 Chr.1: 162374586-162374586 on Build GRCh38
rs548926408 Chr.1: 162374416-162374416 on Build GRCh38
rs1052290 Chr.1: 162374414-162374414 on Build GRCh38
rs776059118 Chr.1: 162374607-162374607 on Build GRCh38
rs774063068 Chr.1: 162374495-162374495 on Build GRCh38
rs372388707 Chr.1: 162374592-162374592 on Build GRCh38
rs770728820 Chr.1: 162374493-162374493 on Build GRCh38
rs1038582902 Chr.1: 162374486-162374486 on Build GRCh38
rs750974852 Chr.1: 162374528-162374528 on Build GRCh38
rs780318701 Chr.1: 162374448-162374448 on Build GRCh38
rs748189838 Chr.1: 162374559-162374559 on Build GRCh38
rs750699614 Chr.1: 162374426-162374426 on Build GRCh38
rs142986032 Chr.1: 162374415-162374415 on Build GRCh38
rs372797714 Chr.1: 162374574-162374574 on Build GRCh38
rs765688142 Chr.1: 162374518-162374518 on Build GRCh38
rs773222858 Chr.1: 162374480-162374480 on Build GRCh38
rs756078533 Chr.1: 162374527-162374527 on Build GRCh38
rs753025600 Chr.1: 162374513-162374513 on Build GRCh38
rs151080283 Chr.1: 162374499-162374499 on Build GRCh38
rs991432455 Chr.1: 162374602-162374602 on Build GRCh38
rs761349149 Chr.1: 162374477-162374483 on Build GRCh38
rs766797888 Chr.1: 162374619-162374619 on Build GRCh38
rs766749186 Chr.1: 162374543-162374543 on Build GRCh38
rs779401029 Chr.1: 162374425-162374425 on Build GRCh38
rs164159 Chr.1: 162374600-162374600 on Build GRCh38
rs891308394 Chr.1: 162374558-162374558 on Build GRCh38
rs557505859 Chr.1: 162374467-162374467 on Build GRCh38
rs751257400 Chr.1: 162374401-162374401 on Build GRCh38
rs767226461 Chr.1: 162374500-162374500 on Build GRCh38
rs765113492 Chr.1: 162374556-162374558 on Build GRCh38
rs758874828 Chr.1: 162374541-162374541 on Build GRCh38
rs982559043 Chr.1: 162374434-162374434 on Build GRCh38
rs941071138 Chr.1: 162374485-162374485 on Build GRCh38
rs138396979 Chr.1: 162374575-162374575 on Build GRCh38
rs182178991 Chr.1: 162374591-162374591 on Build GRCh38
rs374328687 Chr.1: 162374459-162374459 on Build GRCh38
rs370833117 Chr.1: 162374420-162374420 on Build GRCh38
rs1051677146 Chr.1: 162374540-162374540 on Build GRCh38
rs767713166 Chr.1: 162374504-162374504 on Build GRCh38
rs371340536 Chr.1: 162374590-162374590 on Build GRCh38
rs917105228 Chr.1: 162374421-162374421 on Build GRCh38
rs140944179 Chr.1: 162374557-162374557 on Build GRCh38
rs146452310 Chr.1: 162374556-162374556 on Build GRCh38
rs758817112 Chr.1: 162374430-162374430 on Build GRCh38
rs949808395 Chr.1: 162374431-162374431 on Build GRCh38
rs762614616 Chr.1: 162374487-162374487 on Build GRCh38
rs367845900 Chr.1: 162374585-162374585 on Build GRCh38
rs751933453 Chr.1: 162374544-162374544 on Build GRCh38
rs201416465 Chr.1: 162374587-162374587 on Build GRCh38

View Full Product Details