Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 160419056-160419297 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VANGL2
Entrez Gene ID
57216
Gene Name
VANGL planar cell polarity protein 2
Gene Aliases
LPP1, LTAP, STB1, STBM, STBM1
UniGene
Hs.99477
Gene Chromosome Location
Chr.1: 160400574-160428678 on Build GRCh38
Transcripts
NM_020335.2 XM_005245357.1 XM_011509804.1 XM_011509805.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1747867 c.370C>G p.L124V Chr.1: 160419179-160419179 on Build GRCh38
7178010 c.273C>T p.I91I Chr.1: 160419082-160419082 on Build GRCh38
1747866 c.299C>G p.P100R Chr.1: 160419108-160419108 on Build GRCh38
6122243 c.395T>A p.L132Q Chr.1: 160419204-160419204 on Build GRCh38
424328 c.423C>T p.C141C Chr.1: 160419232-160419232 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746867544 Chr.1: 160419219-160419219 on Build GRCh38
rs772529847 Chr.1: 160419244-160419244 on Build GRCh38
rs769374648 Chr.1: 160419238-160419238 on Build GRCh38
rs201184424 Chr.1: 160419266-160419266 on Build GRCh38
rs779651925 Chr.1: 160419128-160419128 on Build GRCh38
rs781329666 Chr.1: 160419236-160419236 on Build GRCh38
rs748048733 Chr.1: 160419237-160419237 on Build GRCh38
rs565547219 Chr.1: 160419130-160419130 on Build GRCh38
rs752999206 Chr.1: 160419191-160419191 on Build GRCh38
rs990144574 Chr.1: 160419173-160419173 on Build GRCh38
rs762473721 Chr.1: 160419172-160419172 on Build GRCh38
rs764417088 Chr.1: 160419277-160419277 on Build GRCh38
rs757382979 Chr.1: 160419212-160419212 on Build GRCh38
rs754790677 Chr.1: 160419234-160419234 on Build GRCh38
rs767724959 Chr.1: 160419082-160419082 on Build GRCh38
rs767929095 Chr.1: 160419175-160419175 on Build GRCh38
rs779232954 Chr.1: 160419119-160419119 on Build GRCh38
rs1057118359 Chr.1: 160419233-160419233 on Build GRCh38
rs746031069 Chr.1: 160419122-160419122 on Build GRCh38
rs759993919 Chr.1: 160419078-160419078 on Build GRCh38
rs199623968 Chr.1: 160419148-160419148 on Build GRCh38
rs753974787 Chr.1: 160419111-160419111 on Build GRCh38
rs757320101 Chr.1: 160419117-160419117 on Build GRCh38
rs563261116 Chr.1: 160419253-160419253 on Build GRCh38
rs756546116 Chr.1: 160419085-160419085 on Build GRCh38
rs940748787 Chr.1: 160419271-160419271 on Build GRCh38
rs149889263 Chr.1: 160419077-160419077 on Build GRCh38
rs200356076 Chr.1: 160419142-160419142 on Build GRCh38
rs761011149 Chr.1: 160419196-160419196 on Build GRCh38
rs1049148642 Chr.1: 160419184-160419184 on Build GRCh38
rs764650174 Chr.1: 160419203-160419203 on Build GRCh38
rs371082262 Chr.1: 160419096-160419096 on Build GRCh38
rs866058743 Chr.1: 160419245-160419245 on Build GRCh38
rs373536899 Chr.1: 160419123-160419123 on Build GRCh38
rs770544019 Chr.1: 160419265-160419265 on Build GRCh38
rs370160195 Chr.1: 160419213-160419213 on Build GRCh38
rs142604710 Chr.1: 160419274-160419274 on Build GRCh38
rs761450874 Chr.1: 160419141-160419141 on Build GRCh38
rs780411852 Chr.1: 160419216-160419216 on Build GRCh38
rs754308820 Chr.1: 160419208-160419208 on Build GRCh38
rs376432810 Chr.1: 160419132-160419132 on Build GRCh38
rs140250013 Chr.1: 160419083-160419083 on Build GRCh38

View Full Product Details