Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 33086921-33087192 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSTF3
Entrez Gene ID
1479
Gene Name
cleavage stimulation factor subunit 3
Gene Aliases
CSTF-77
UniGene
Hs.44402
Gene Chromosome Location
Chr.11: 33084584-33161491 on Build GRCh38
Transcripts
NM_001326.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6132503 c.1720G>A p.E574K Chr.11: 33087063-33087063 on Build GRCh38
7296496 c.1795+1G>A p.? Chr.11: 33086987-33086987 on Build GRCh38
4032436 c.1783C>T p.R595* Chr.11: 33087000-33087000 on Build GRCh38
7251950 c.1754A>G p.D585G Chr.11: 33087029-33087029 on Build GRCh38
6637062 c.1784_1786delGAC p.R595delR Chr.11: 33086997-33086999 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764581305 Chr.11: 33087033-33087033 on Build GRCh38
rs775825908 Chr.11: 33087011-33087011 on Build GRCh38
rs765914953 Chr.11: 33087044-33087044 on Build GRCh38
rs769717692 Chr.11: 33087123-33087123 on Build GRCh38
rs147624640 Chr.11: 33087106-33087106 on Build GRCh38
rs762417208 Chr.11: 33087041-33087041 on Build GRCh38
rs377006746 Chr.11: 33087055-33087055 on Build GRCh38
rs760913649 Chr.11: 33087024-33087024 on Build GRCh38
rs779541503 Chr.11: 33087064-33087064 on Build GRCh38
rs774417141 Chr.11: 33086961-33086961 on Build GRCh38
rs754345078 Chr.11: 33087034-33087034 on Build GRCh38
rs749891940 Chr.11: 33087054-33087054 on Build GRCh38
rs754720144 Chr.11: 33087098-33087098 on Build GRCh38
rs772171033 Chr.11: 33086998-33086998 on Build GRCh38
rs747685177 Chr.11: 33086952-33086954 on Build GRCh38
rs1802709 Chr.11: 33087122-33087122 on Build GRCh38
rs777824622 Chr.11: 33087145-33087145 on Build GRCh38
rs759728029 Chr.11: 33086980-33086980 on Build GRCh38
rs192870893 Chr.11: 33087149-33087149 on Build GRCh38
rs756792158 Chr.11: 33087072-33087072 on Build GRCh38
rs772229084 Chr.11: 33087157-33087157 on Build GRCh38
rs748852024 Chr.11: 33087161-33087173 on Build GRCh38
rs576868118 Chr.11: 33086948-33086948 on Build GRCh38
rs910314397 Chr.11: 33087114-33087114 on Build GRCh38
rs994110520 Chr.11: 33086955-33086955 on Build GRCh38
rs770938119 Chr.11: 33086956-33086956 on Build GRCh38
rs745985559 Chr.11: 33087084-33087084 on Build GRCh38
rs768536528 Chr.11: 33086951-33086951 on Build GRCh38
rs1025957959 Chr.11: 33086986-33086986 on Build GRCh38
rs868783384 Chr.11: 33087133-33087133 on Build GRCh38
rs202158353 Chr.11: 33086954-33086954 on Build GRCh38
rs201667196 Chr.11: 33086953-33086953 on Build GRCh38
rs748010913 Chr.11: 33087107-33087107 on Build GRCh38

View Full Product Details