Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 83915050-83915321 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MLYCD
Entrez Gene ID
23417
Gene Name
malonyl-CoA decarboxylase
Gene Aliases
MCD
UniGene
Hs.644610
Gene Chromosome Location
Chr.16: 83899125-83916182 on Build GRCh38
Transcripts
NM_012213.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4459651 c.1134C>T p.L378L Chr.16: 83915141-83915141 on Build GRCh38
3717111 c.1175G>T p.R392L Chr.16: 83915182-83915182 on Build GRCh38
380409 c.1227G>T p.E409D Chr.16: 83915234-83915234 on Build GRCh38
974300 c.1073C>T p.S358L Chr.16: 83915080-83915080 on Build GRCh38
6364790 c.1288C>A p.L430M Chr.16: 83915295-83915295 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372255821 Chr.16: 83915250-83915250 on Build GRCh38
rs752642605 Chr.16: 83915246-83915246 on Build GRCh38
rs368099850 Chr.16: 83915248-83915248 on Build GRCh38
rs772112058 Chr.16: 83915223-83915223 on Build GRCh38
rs886202991 Chr.16: 83915115-83915115 on Build GRCh38
rs201973830 Chr.16: 83915302-83915302 on Build GRCh38
rs200573073 Chr.16: 83915080-83915080 on Build GRCh38
rs751649669 Chr.16: 83915129-83915129 on Build GRCh38
rs775795977 Chr.16: 83915285-83915286 on Build GRCh38
rs35124955 Chr.16: 83915301-83915301 on Build GRCh38
rs776015473 Chr.16: 83915124-83915124 on Build GRCh38
rs780327700 Chr.16: 83915211-83915211 on Build GRCh38
rs375981938 Chr.16: 83915295-83915295 on Build GRCh38
rs759839871 Chr.16: 83915116-83915116 on Build GRCh38
rs995534057 Chr.16: 83915150-83915150 on Build GRCh38
rs771229830 Chr.16: 83915283-83915283 on Build GRCh38
rs200750439 Chr.16: 83915185-83915185 on Build GRCh38
rs368357572 Chr.16: 83915274-83915274 on Build GRCh38
rs756042120 Chr.16: 83915093-83915093 on Build GRCh38
rs369287252 Chr.16: 83915297-83915297 on Build GRCh38
rs749679659 Chr.16: 83915213-83915213 on Build GRCh38
rs377161125 Chr.16: 83915081-83915081 on Build GRCh38
rs528963936 Chr.16: 83915267-83915267 on Build GRCh38
rs772187327 Chr.16: 83915219-83915220 on Build GRCh38
rs1037462236 Chr.16: 83915253-83915253 on Build GRCh38
rs767761325 Chr.16: 83915187-83915187 on Build GRCh38
rs375614850 Chr.16: 83915261-83915261 on Build GRCh38
rs762025589 Chr.16: 83915131-83915131 on Build GRCh38
rs762729042 Chr.16: 83915288-83915288 on Build GRCh38
rs770311684 Chr.16: 83915123-83915123 on Build GRCh38
rs753701525 Chr.16: 83915198-83915198 on Build GRCh38
rs372909321 Chr.16: 83915096-83915096 on Build GRCh38
rs748788635 Chr.16: 83915095-83915095 on Build GRCh38
rs3803646 Chr.16: 83915085-83915085 on Build GRCh38
rs200341673 Chr.16: 83915210-83915210 on Build GRCh38
rs202210941 Chr.16: 83915154-83915154 on Build GRCh38
rs750654348 Chr.16: 83915193-83915193 on Build GRCh38
rs530627828 Chr.16: 83915181-83915181 on Build GRCh38
rs748231506 Chr.16: 83915221-83915221 on Build GRCh38
rs569549800 Chr.16: 83915214-83915214 on Build GRCh38
rs750981358 Chr.16: 83915292-83915292 on Build GRCh38
rs750187634 Chr.16: 83915141-83915141 on Build GRCh38
rs757910788 Chr.16: 83915247-83915247 on Build GRCh38
rs773101554 Chr.16: 83915262-83915262 on Build GRCh38
rs767659723 Chr.16: 83915140-83915140 on Build GRCh38
rs747617187 Chr.16: 83915160-83915160 on Build GRCh38
rs765693763 Chr.16: 83915074-83915074 on Build GRCh38
rs766715091 Chr.16: 83915238-83915238 on Build GRCh38
rs1039004103 Chr.16: 83915098-83915098 on Build GRCh38
rs753847742 Chr.16: 83915158-83915158 on Build GRCh38
rs760517323 Chr.16: 83915196-83915196 on Build GRCh38
rs577969223 Chr.16: 83915077-83915077 on Build GRCh38
rs777475399 Chr.16: 83915207-83915207 on Build GRCh38
rs776527401 Chr.16: 83915240-83915240 on Build GRCh38
rs780166693 Chr.16: 83915094-83915094 on Build GRCh38
rs760752300 Chr.16: 83915229-83915229 on Build GRCh38
rs374610037 Chr.16: 83915206-83915206 on Build GRCh38
rs373273203 Chr.16: 83915153-83915153 on Build GRCh38
rs371429195 Chr.16: 83915290-83915290 on Build GRCh38
rs181840735 Chr.16: 83915099-83915099 on Build GRCh38
rs745651086 Chr.16: 83915092-83915092 on Build GRCh38
rs766336109 Chr.16: 83915197-83915197 on Build GRCh38
rs779026116 Chr.16: 83915200-83915200 on Build GRCh38
rs755915608 Chr.16: 83915142-83915142 on Build GRCh38
rs561510435 Chr.16: 83915168-83915168 on Build GRCh38
rs777032506 Chr.16: 83915284-83915284 on Build GRCh38
rs779473261 Chr.16: 83915256-83915256 on Build GRCh38
rs763014509 Chr.16: 83915126-83915126 on Build GRCh38
rs746697130 Chr.16: 83915209-83915209 on Build GRCh38
rs199549095 Chr.16: 83915105-83915105 on Build GRCh38
rs370278897 Chr.16: 83915243-83915243 on Build GRCh38
rs758239547 Chr.16: 83915252-83915252 on Build GRCh38
rs201576638 Chr.16: 83915182-83915182 on Build GRCh38
rs564269563 Chr.16: 83915186-83915186 on Build GRCh38
rs746164922 Chr.16: 83915186-83915186 on Build GRCh38
rs376324546 Chr.16: 83915167-83915167 on Build GRCh38
rs369896830 Chr.16: 83915090-83915090 on Build GRCh38
rs374608119 Chr.16: 83915244-83915244 on Build GRCh38
rs377271580 Chr.16: 83915286-83915286 on Build GRCh38
rs752228353 Chr.16: 83915201-83915201 on Build GRCh38
rs375649018 Chr.16: 83915227-83915227 on Build GRCh38
rs747868539 Chr.16: 83915102-83915102 on Build GRCh38
rs367874845 Chr.16: 83915285-83915285 on Build GRCh38
rs376504760 Chr.16: 83915104-83915104 on Build GRCh38
rs764214677 Chr.16: 83915128-83915128 on Build GRCh38
rs751083468 Chr.16: 83915249-83915249 on Build GRCh38
rs774773160 Chr.16: 83915216-83915216 on Build GRCh38

View Full Product Details