Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 59338676-59338907 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LMAN1
Entrez Gene ID
3998
Gene Name
lectin, mannose binding 1
Gene Aliases
ERGIC-53, ERGIC53, F5F8D, FMFD1, MCFD1, MR60, gp58
UniGene
Hs.465295
Gene Chromosome Location
Chr.18: 59327823-59359276 on Build GRCh38
Transcripts
NM_005570.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4802224 c.1114A>G p.K372E Chr.18: 59338795-59338795 on Build GRCh38
4995813 c.1091C>T p.S364F Chr.18: 59338818-59338818 on Build GRCh38
3970600 c.1018C>T p.R340C Chr.18: 59338891-59338891 on Build GRCh38
3821626 c.1020T>C p.R340R Chr.18: 59338889-59338889 on Build GRCh38
989341 c.1051C>T p.Q351* Chr.18: 59338858-59338858 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1021076846 Chr.18: 59338840-59338840 on Build GRCh38
rs375670001 Chr.18: 59338778-59338778 on Build GRCh38
rs763218719 Chr.18: 59338729-59338729 on Build GRCh38
rs555831589 Chr.18: 59338733-59338733 on Build GRCh38
rs3737392 Chr.18: 59338845-59338845 on Build GRCh38
rs181218003 Chr.18: 59338821-59338821 on Build GRCh38
rs185875271 Chr.18: 59338859-59338859 on Build GRCh38
rs764842259 Chr.18: 59338822-59338822 on Build GRCh38
rs766591385 Chr.18: 59338801-59338801 on Build GRCh38
rs746732718 Chr.18: 59338861-59338861 on Build GRCh38
rs768125559 Chr.18: 59338773-59338773 on Build GRCh38
rs761961117 Chr.18: 59338742-59338742 on Build GRCh38
rs891524949 Chr.18: 59338771-59338771 on Build GRCh38
rs1040811142 Chr.18: 59338722-59338722 on Build GRCh38
rs367976630 Chr.18: 59338823-59338823 on Build GRCh38
rs768245714 Chr.18: 59338768-59338776 on Build GRCh38
rs777835065 Chr.18: 59338848-59338848 on Build GRCh38
rs78116104 Chr.18: 59338855-59338855 on Build GRCh38
rs757096830 Chr.18: 59338860-59338860 on Build GRCh38
rs567534318 Chr.18: 59338834-59338834 on Build GRCh38
rs970097793 Chr.18: 59338827-59338827 on Build GRCh38
rs869312031 Chr.18: 59338758-59338758 on Build GRCh38
rs753515236 Chr.18: 59338813-59338813 on Build GRCh38
rs1008050264 Chr.18: 59338728-59338728 on Build GRCh38
rs780008620 Chr.18: 59338739-59338739 on Build GRCh38
rs1011664151 Chr.18: 59338819-59338819 on Build GRCh38
rs1022011064 Chr.18: 59338826-59338826 on Build GRCh38
rs199873517 Chr.18: 59338716-59338716 on Build GRCh38
rs41358147 Chr.18: 59338723-59338723 on Build GRCh38

View Full Product Details