Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2206996-2207253 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MLST8
Entrez Gene ID
64223
Gene Name
MTOR associated protein, LST8 homolog
Gene Aliases
GBL, GbetaL, LST8, POP3, WAT1
UniGene
Hs.29203
Gene Chromosome Location
Chr.16: 2205177-2209417 on Build GRCh38
Transcripts
NM_001199173.1 NM_001199174.1 NM_001199175.1 NM_022372.4 XM_005255475.3 XM_005255478.2 XM_005255479.2 XM_017023548.1 XM_017023549.1 XM_017023550.1 XM_017023551.1 XM_017023552.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7275400 c.345-9G>A p.? Chr.16: 2207026-2207026 on Build GRCh38
702275 c.347C>T p.S116F Chr.16: 2207037-2207037 on Build GRCh38
3507682 c.369G>A p.R123R Chr.16: 2207059-2207059 on Build GRCh38
3754791 c.414C>G p.P138P Chr.16: 2207104-2207104 on Build GRCh38
3507685 c.442C>T p.Q148* Chr.16: 2207214-2207214 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770986995 Chr.16: 2207207-2207207 on Build GRCh38
rs766345206 Chr.16: 2207137-2207137 on Build GRCh38
rs970322128 Chr.16: 2207098-2207098 on Build GRCh38
rs774698942 Chr.16: 2207120-2207120 on Build GRCh38
rs776018846 Chr.16: 2207020-2207020 on Build GRCh38
rs189570765 Chr.16: 2207050-2207050 on Build GRCh38
rs764920142 Chr.16: 2207058-2207058 on Build GRCh38
rs769036084 Chr.16: 2207024-2207024 on Build GRCh38
rs751067672 Chr.16: 2207168-2207168 on Build GRCh38
rs377624870 Chr.16: 2207226-2207226 on Build GRCh38
rs775982928 Chr.16: 2207086-2207086 on Build GRCh38
rs747348355 Chr.16: 2207157-2207157 on Build GRCh38
rs374170484 Chr.16: 2207219-2207219 on Build GRCh38
rs772218346 Chr.16: 2207188-2207188 on Build GRCh38
rs776528027 Chr.16: 2207141-2207141 on Build GRCh38
rs780777589 Chr.16: 2207163-2207163 on Build GRCh38
rs369716099 Chr.16: 2207185-2207185 on Build GRCh38
rs746364035 Chr.16: 2207014-2207014 on Build GRCh38
rs749626779 Chr.16: 2207021-2207021 on Build GRCh38
rs984225736 Chr.16: 2207175-2207175 on Build GRCh38
rs372872486 Chr.16: 2207164-2207164 on Build GRCh38
rs26862 Chr.16: 2207104-2207104 on Build GRCh38
rs780671810 Chr.16: 2207101-2207101 on Build GRCh38
rs759386473 Chr.16: 2207144-2207144 on Build GRCh38
rs748326365 Chr.16: 2207125-2207125 on Build GRCh38
rs746479398 Chr.16: 2207089-2207089 on Build GRCh38
rs747141800 Chr.16: 2207204-2207204 on Build GRCh38
rs780593582 Chr.16: 2207146-2207146 on Build GRCh38
rs773191686 Chr.16: 2207134-2207134 on Build GRCh38
rs750874624 Chr.16: 2207017-2207018 on Build GRCh38
rs904410434 Chr.16: 2207225-2207225 on Build GRCh38
rs780761564 Chr.16: 2207180-2207180 on Build GRCh38
rs778026039 Chr.16: 2207194-2207194 on Build GRCh38
rs148259622 Chr.16: 2207170-2207170 on Build GRCh38
rs200537646 Chr.16: 2207148-2207148 on Build GRCh38
rs758000683 Chr.16: 2207074-2207074 on Build GRCh38
rs901138450 Chr.16: 2207026-2207026 on Build GRCh38
rs762007437 Chr.16: 2207028-2207028 on Build GRCh38
rs779162509 Chr.16: 2207183-2207183 on Build GRCh38
rs930748420 Chr.16: 2207017-2207017 on Build GRCh38
rs760709050 Chr.16: 2207135-2207135 on Build GRCh38
rs185496759 Chr.16: 2207030-2207030 on Build GRCh38
rs374173728 Chr.16: 2207069-2207069 on Build GRCh38
rs753665679 Chr.16: 2207045-2207045 on Build GRCh38
rs978979894 Chr.16: 2207057-2207057 on Build GRCh38
rs774412241 Chr.16: 2207025-2207025 on Build GRCh38
rs760546991 Chr.16: 2207039-2207039 on Build GRCh38
rs756720316 Chr.16: 2207094-2207094 on Build GRCh38
rs763761876 Chr.16: 2207165-2207165 on Build GRCh38
rs28124 Chr.16: 2207152-2207152 on Build GRCh38
rs772282045 Chr.16: 2207126-2207126 on Build GRCh38
rs770367150 Chr.16: 2207018-2207018 on Build GRCh38
rs749806563 Chr.16: 2207181-2207181 on Build GRCh38
rs992540291 Chr.16: 2207088-2207088 on Build GRCh38
rs368472438 Chr.16: 2207031-2207031 on Build GRCh38
rs180952140 Chr.16: 2207145-2207145 on Build GRCh38
rs766259271 Chr.16: 2207040-2207040 on Build GRCh38
rs755372927 Chr.16: 2207160-2207161 on Build GRCh38
rs758823938 Chr.16: 2207115-2207116 on Build GRCh38
rs752477490 Chr.16: 2207160-2207160 on Build GRCh38
rs900500229 Chr.16: 2207190-2207190 on Build GRCh38
rs371612671 Chr.16: 2207117-2207117 on Build GRCh38

View Full Product Details