Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6967893-6968120 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHB2
Entrez Gene ID
11331
Gene Name
prohibitin 2
Gene Aliases
BAP, BCAP37, Bap37, PNAS-141, REA, hBAP, p22
UniGene
Hs.504620
Gene Chromosome Location
Chr.12: 6965352-6970823 on Build GRCh38
Transcripts
NM_001144831.1 NM_001267700.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5421007 c.538G>T p.D180Y Chr.12: 6967961-6967961 on Build GRCh38
6744827 c.493C>T p.R165C Chr.12: 6968006-6968006 on Build GRCh38
4875099 c.554C>T p.T185I Chr.12: 6967945-6967945 on Build GRCh38
5650234 c.510G>A p.E170E Chr.12: 6967989-6967989 on Build GRCh38
6073723 c.484C>T p.L162L Chr.12: 6968015-6968015 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782352200 Chr.12: 6968029-6968029 on Build GRCh38
rs781990930 Chr.12: 6967928-6967928 on Build GRCh38
rs367716554 Chr.12: 6967998-6967998 on Build GRCh38
rs948890705 Chr.12: 6967999-6967999 on Build GRCh38
rs782294470 Chr.12: 6968065-6968065 on Build GRCh38
rs782623339 Chr.12: 6968052-6968052 on Build GRCh38
rs782558395 Chr.12: 6968032-6968032 on Build GRCh38
rs762573405 Chr.12: 6968094-6968094 on Build GRCh38
rs375814822 Chr.12: 6968044-6968044 on Build GRCh38
rs781943177 Chr.12: 6967919-6967919 on Build GRCh38
rs782385638 Chr.12: 6968056-6968056 on Build GRCh38
rs782674765 Chr.12: 6968064-6968064 on Build GRCh38
rs1031648105 Chr.12: 6967926-6967926 on Build GRCh38
rs782786778 Chr.12: 6967985-6967985 on Build GRCh38
rs782503992 Chr.12: 6968026-6968026 on Build GRCh38
rs782555915 Chr.12: 6968043-6968043 on Build GRCh38
rs371492548 Chr.12: 6967927-6967927 on Build GRCh38
rs782253805 Chr.12: 6968053-6968053 on Build GRCh38
rs782585743 Chr.12: 6968034-6968034 on Build GRCh38
rs782312878 Chr.12: 6968069-6968069 on Build GRCh38
rs782728839 Chr.12: 6968006-6968006 on Build GRCh38
rs781839686 Chr.12: 6968018-6968018 on Build GRCh38
rs782221432 Chr.12: 6967920-6967920 on Build GRCh38
rs376134268 Chr.12: 6967995-6967995 on Build GRCh38
rs371919995 Chr.12: 6968009-6968009 on Build GRCh38
rs782131423 Chr.12: 6967949-6967949 on Build GRCh38
rs782549493 Chr.12: 6968051-6968051 on Build GRCh38

View Full Product Details