Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32191526-32191759 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPSM3
Entrez Gene ID
63940
Gene Name
G-protein signaling modulator 3
Gene Aliases
AGS4, C6orf9, G18, G18.1a, G18.1b, G18.2, NG1
UniGene
Hs.520046
Gene Chromosome Location
Chr.6: 32190766-32195523 on Build GRCh38
Transcripts
NM_001276501.1 NM_022107.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3872954 c.335T>A p.L112H Chr.6: 32191719-32191719 on Build GRCh38
3872955 c.296G>A p.R99H Chr.6: 32191758-32191758 on Build GRCh38
3625359 c.343C>A p.Q115K Chr.6: 32191711-32191711 on Build GRCh38
6674696 c.327C>G p.S109R Chr.6: 32191727-32191727 on Build GRCh38
3394112 c.303C>T p.L101L Chr.6: 32191751-32191751 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759230089 Chr.6: 32191620-32191620 on Build GRCh38
rs1041971758 Chr.6: 32191717-32191718 on Build GRCh38
rs529089304 Chr.6: 32191626-32191626 on Build GRCh38
rs926324696 Chr.6: 32191656-32191656 on Build GRCh38
rs776463249 Chr.6: 32191683-32191683 on Build GRCh38
rs757940985 Chr.6: 32191671-32191671 on Build GRCh38
rs903458508 Chr.6: 32191723-32191723 on Build GRCh38
rs997051686 Chr.6: 32191585-32191585 on Build GRCh38
rs775017295 Chr.6: 32191704-32191704 on Build GRCh38
rs970789750 Chr.6: 32191641-32191641 on Build GRCh38
rs991626425 Chr.6: 32191662-32191662 on Build GRCh38
rs1055431632 Chr.6: 32191643-32191644 on Build GRCh38
rs950194790 Chr.6: 32191716-32191716 on Build GRCh38
rs1026790229 Chr.6: 32191588-32191588 on Build GRCh38
rs987393144 Chr.6: 32191575-32191575 on Build GRCh38
rs746677036 Chr.6: 32191676-32191676 on Build GRCh38
rs973507650 Chr.6: 32191602-32191602 on Build GRCh38
rs540132973 Chr.6: 32191606-32191606 on Build GRCh38
rs770686786 Chr.6: 32191681-32191681 on Build GRCh38
rs879423067 Chr.6: 32191612-32191612 on Build GRCh38
rs747709742 Chr.6: 32191663-32191663 on Build GRCh38
rs777510082 Chr.6: 32191675-32191675 on Build GRCh38
rs952605990 Chr.6: 32191596-32191596 on Build GRCh38
rs771545085 Chr.6: 32191579-32191579 on Build GRCh38
rs561910133 Chr.6: 32191611-32191611 on Build GRCh38
rs879606937 Chr.6: 32191595-32191595 on Build GRCh38
rs1015491824 Chr.6: 32191621-32191621 on Build GRCh38
rs745800137 Chr.6: 32191696-32191696 on Build GRCh38
rs544726553 Chr.6: 32191660-32191660 on Build GRCh38
rs981839642 Chr.6: 32191649-32191649 on Build GRCh38
rs769726202 Chr.6: 32191697-32191697 on Build GRCh38

View Full Product Details