Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 74882481-74882746 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FADS6
Entrez Gene ID
283985
Gene Name
fatty acid desaturase 6
Gene Aliases
FP18279
UniGene
Hs.448845
Gene Chromosome Location
Chr.17: 74874793-74893811 on Build GRCh38
Transcripts
NM_178128.5 XM_005257224.4 XM_017024458.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5544933 c.511C>T p.P171S Chr.17: 74882557-74882557 on Build GRCh38
7202003 c.390G>T p.T130T Chr.17: 74882678-74882678 on Build GRCh38
4548171 c.440G>A p.G147E Chr.17: 74882628-74882628 on Build GRCh38
3521959 c.368C>T p.A123V Chr.17: 74882700-74882700 on Build GRCh38
3521957 c.441G>A p.G147G Chr.17: 74882627-74882627 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755683985 Chr.17: 74882642-74882642 on Build GRCh38
rs369319580 Chr.17: 74882684-74882684 on Build GRCh38
rs938109344 Chr.17: 74882616-74882616 on Build GRCh38
rs769642366 Chr.17: 74882579-74882579 on Build GRCh38
rs749255006 Chr.17: 74882581-74882581 on Build GRCh38
rs377760070 Chr.17: 74882531-74882531 on Build GRCh38
rs747743130 Chr.17: 74882574-74882574 on Build GRCh38
rs371257165 Chr.17: 74882552-74882552 on Build GRCh38
rs753979983 Chr.17: 74882530-74882530 on Build GRCh38
rs755628485 Chr.17: 74882724-74882724 on Build GRCh38
rs367741660 Chr.17: 74882670-74882670 on Build GRCh38
rs934914317 Chr.17: 74882691-74882691 on Build GRCh38
rs757016071 Chr.17: 74882653-74882653 on Build GRCh38
rs767030840 Chr.17: 74882615-74882615 on Build GRCh38
rs560453575 Chr.17: 74882610-74882610 on Build GRCh38
rs374233442 Chr.17: 74882558-74882558 on Build GRCh38
rs931115552 Chr.17: 74882585-74882585 on Build GRCh38
rs568041144 Chr.17: 74882566-74882566 on Build GRCh38
rs775510370 Chr.17: 74882520-74882520 on Build GRCh38
rs775010554 Chr.17: 74882689-74882689 on Build GRCh38
rs377636516 Chr.17: 74882569-74882569 on Build GRCh38
rs779636351 Chr.17: 74882502-74882502 on Build GRCh38
rs766880524 Chr.17: 74882639-74882639 on Build GRCh38
rs760909285 Chr.17: 74882522-74882522 on Build GRCh38
rs768301326 Chr.17: 74882505-74882505 on Build GRCh38
rs577982165 Chr.17: 74882596-74882596 on Build GRCh38
rs772493939 Chr.17: 74882506-74882506 on Build GRCh38
rs115820690 Chr.17: 74882583-74882583 on Build GRCh38
rs963069234 Chr.17: 74882627-74882627 on Build GRCh38
rs141147037 Chr.17: 74882580-74882580 on Build GRCh38
rs774251462 Chr.17: 74882592-74882592 on Build GRCh38
rs374540498 Chr.17: 74882654-74882654 on Build GRCh38
rs745782129 Chr.17: 74882668-74882668 on Build GRCh38
rs763778782 Chr.17: 74882725-74882725 on Build GRCh38
rs373453144 Chr.17: 74882528-74882528 on Build GRCh38
rs759892994 Chr.17: 74882696-74882696 on Build GRCh38
rs765509140 Chr.17: 74882542-74882542 on Build GRCh38
rs748421271 Chr.17: 74882672-74882672 on Build GRCh38
rs752398046 Chr.17: 74882641-74882641 on Build GRCh38
rs368990242 Chr.17: 74882511-74882511 on Build GRCh38
rs778891818 Chr.17: 74882660-74882660 on Build GRCh38
rs763385048 Chr.17: 74882612-74882612 on Build GRCh38
rs772145870 Chr.17: 74882516-74882516 on Build GRCh38
rs770185420 Chr.17: 74882678-74882678 on Build GRCh38
rs146475710 Chr.17: 74882510-74882510 on Build GRCh38
rs753486346 Chr.17: 74882726-74882726 on Build GRCh38
rs763475295 Chr.17: 74882681-74882681 on Build GRCh38
rs574003859 Chr.17: 74882679-74882679 on Build GRCh38
rs746687655 Chr.17: 74882503-74882503 on Build GRCh38
rs750983186 Chr.17: 74882545-74882545 on Build GRCh38
rs748014587 Chr.17: 74882508-74882508 on Build GRCh38
rs754572438 Chr.17: 74882617-74882617 on Build GRCh38
rs896437877 Chr.17: 74882645-74882645 on Build GRCh38
rs768616673 Chr.17: 74882594-74882594 on Build GRCh38
rs540230751 Chr.17: 74882601-74882601 on Build GRCh38
rs752100935 Chr.17: 74882715-74882715 on Build GRCh38
rs889530194 Chr.17: 74882703-74882703 on Build GRCh38
rs949419206 Chr.17: 74882657-74882657 on Build GRCh38
rs777568064 Chr.17: 74882643-74882643 on Build GRCh38
rs749132308 Chr.17: 74882646-74882646 on Build GRCh38
rs747154502 Chr.17: 74882593-74882593 on Build GRCh38
rs767084686 Chr.17: 74882706-74882706 on Build GRCh38
rs566760752 Chr.17: 74882547-74882547 on Build GRCh38

View Full Product Details