Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 87163290-87163561 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLHL8
Entrez Gene ID
57563
Gene Name
kelch like family member 8
Gene Aliases
-
UniGene
Hs.106601
Gene Chromosome Location
Chr.4: 87160103-87220608 on Build GRCh38
Transcripts
NM_001292003.1 NM_001292006.1 NM_001292007.1 NM_020803.4 NR_120447.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1722160 c.1839C>T p.S613S Chr.4: 87163543-87163543 on Build GRCh38
5737018 c.1850T>C p.V617A Chr.4: 87163532-87163532 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs542435648 Chr.4: 87163363-87163363 on Build GRCh38
rs145974248 Chr.4: 87163520-87163520 on Build GRCh38
rs931936691 Chr.4: 87163478-87163478 on Build GRCh38
rs576394937 Chr.4: 87163405-87163405 on Build GRCh38
rs768896779 Chr.4: 87163497-87163497 on Build GRCh38
rs760767003 Chr.4: 87163508-87163508 on Build GRCh38
rs754112069 Chr.4: 87163513-87163513 on Build GRCh38
rs774960737 Chr.4: 87163499-87163499 on Build GRCh38
rs766717322 Chr.4: 87163510-87163510 on Build GRCh38
rs990010146 Chr.4: 87163349-87163349 on Build GRCh38
rs768466178 Chr.4: 87163516-87163517 on Build GRCh38
rs780765268 Chr.4: 87163490-87163490 on Build GRCh38
rs371352890 Chr.4: 87163485-87163485 on Build GRCh38
rs770576136 Chr.4: 87163488-87163488 on Build GRCh38
rs762463224 Chr.4: 87163444-87163444 on Build GRCh38
rs749719663 Chr.4: 87163496-87163496 on Build GRCh38
rs747477471 Chr.4: 87163426-87163426 on Build GRCh38
rs754444404 Chr.4: 87163341-87163342 on Build GRCh38
rs906467599 Chr.4: 87163420-87163420 on Build GRCh38
rs762202041 Chr.4: 87163500-87163500 on Build GRCh38
rs1049061594 Chr.4: 87163506-87163506 on Build GRCh38
rs746484134 Chr.4: 87163487-87163487 on Build GRCh38
rs936760714 Chr.4: 87163348-87163348 on Build GRCh38
rs772655204 Chr.4: 87163503-87163503 on Build GRCh38
rs189331725 Chr.4: 87163505-87163505 on Build GRCh38
rs922024015 Chr.4: 87163451-87163451 on Build GRCh38
rs573168621 Chr.4: 87163384-87163384 on Build GRCh38

View Full Product Details