Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 132117243-132117512 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JAKMIP3
Entrez Gene ID
282973
Gene Name
Janus kinase and microtubule interacting protein 3
Gene Aliases
C10orf14, C10orf39, Jamip3, NECC2, bA140A10.5
UniGene
Hs.106254
Gene Chromosome Location
Chr.10: 132036068-132184852 on Build GRCh38
Transcripts
NM_001105521.3 NM_001323086.1 NM_001323087.1 NM_001323088.1 NM_001323089.1 NM_001323090.1 XM_005252674.4 XM_005252676.4 XM_011539676.2 XM_011539682.2 XM_017016095.1 XM_017016096.1 XM_017016097.1 XM_017016098.1 XM_017016099.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4547361 c.420G>A p.K140K Chr.10: 132117361-132117361 on Build GRCh38
379826 c.504G>C p.V168V Chr.10: 132117445-132117445 on Build GRCh38
5904354 c.421G>A p.E141K Chr.10: 132117362-132117362 on Build GRCh38
4543821 c.344G>A p.R115Q Chr.10: 132117285-132117285 on Build GRCh38
5919894 c.391G>A p.V131I Chr.10: 132117332-132117332 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1016145065 Chr.10: 132117444-132117444 on Build GRCh38
rs781090131 Chr.10: 132117362-132117363 on Build GRCh38
rs376695053 Chr.10: 132117419-132117419 on Build GRCh38
rs376645431 Chr.10: 132117454-132117454 on Build GRCh38
rs761693157 Chr.10: 132117312-132117312 on Build GRCh38
rs372132098 Chr.10: 132117341-132117341 on Build GRCh38
rs769498280 Chr.10: 132117332-132117332 on Build GRCh38
rs757728403 Chr.10: 132117327-132117327 on Build GRCh38
rs376174504 Chr.10: 132117334-132117334 on Build GRCh38
rs559504656 Chr.10: 132117301-132117301 on Build GRCh38
rs771919412 Chr.10: 132117442-132117442 on Build GRCh38
rs372347107 Chr.10: 132117306-132117306 on Build GRCh38
rs754442102 Chr.10: 132117491-132117491 on Build GRCh38
rs752743833 Chr.10: 132117375-132117375 on Build GRCh38
rs751886326 Chr.10: 132117382-132117382 on Build GRCh38
rs368574779 Chr.10: 132117296-132117296 on Build GRCh38
rs772009063 Chr.10: 132117344-132117344 on Build GRCh38
rs748789156 Chr.10: 132117429-132117429 on Build GRCh38
rs987535619 Chr.10: 132117309-132117309 on Build GRCh38
rs371369909 Chr.10: 132117274-132117274 on Build GRCh38
rs758624911 Chr.10: 132117325-132117325 on Build GRCh38
rs760600589 Chr.10: 132117355-132117355 on Build GRCh38
rs565537154 Chr.10: 132117377-132117377 on Build GRCh38
rs747360916 Chr.10: 132117326-132117326 on Build GRCh38
rs747898590 Chr.10: 132117436-132117436 on Build GRCh38
rs1046201269 Chr.10: 132117380-132117380 on Build GRCh38
rs373833963 Chr.10: 132117285-132117285 on Build GRCh38
rs773065859 Chr.10: 132117352-132117352 on Build GRCh38
rs931724496 Chr.10: 132117295-132117295 on Build GRCh38
rs371280434 Chr.10: 132117459-132117459 on Build GRCh38
rs368486466 Chr.10: 132117319-132117319 on Build GRCh38
rs747891623 Chr.10: 132117493-132117493 on Build GRCh38
rs767446488 Chr.10: 132117315-132117315 on Build GRCh38
rs780655388 Chr.10: 132117477-132117477 on Build GRCh38
rs200538281 Chr.10: 132117340-132117340 on Build GRCh38
rs375429968 Chr.10: 132117311-132117311 on Build GRCh38
rs1001581323 Chr.10: 132117439-132117439 on Build GRCh38
rs778281162 Chr.10: 132117492-132117492 on Build GRCh38
rs1029094173 Chr.10: 132117284-132117284 on Build GRCh38
rs761617138 Chr.10: 132117343-132117343 on Build GRCh38
rs763116421 Chr.10: 132117376-132117376 on Build GRCh38
rs542760197 Chr.10: 132117293-132117293 on Build GRCh38
rs779747219 Chr.10: 132117424-132117424 on Build GRCh38
rs770817262 Chr.10: 132117449-132117449 on Build GRCh38
rs374511964 Chr.10: 132117443-132117443 on Build GRCh38
rs752983229 Chr.10: 132117320-132117320 on Build GRCh38
rs906682620 Chr.10: 132117358-132117358 on Build GRCh38
rs146722402 Chr.10: 132117467-132117467 on Build GRCh38
rs373128130 Chr.10: 132117399-132117399 on Build GRCh38
rs765358533 Chr.10: 132117356-132117356 on Build GRCh38
rs772360908 Chr.10: 132117275-132117275 on Build GRCh38
rs377151206 Chr.10: 132117469-132117469 on Build GRCh38
rs747520782 Chr.10: 132117266-132117266 on Build GRCh38
rs753298946 Chr.10: 132117484-132117484 on Build GRCh38
rs751879204 Chr.10: 132117460-132117460 on Build GRCh38
rs371619151 Chr.10: 132117331-132117331 on Build GRCh38
rs751033443 Chr.10: 132117418-132117418 on Build GRCh38
rs757577685 Chr.10: 132117383-132117383 on Build GRCh38
rs369892382 Chr.10: 132117430-132117430 on Build GRCh38
rs750781803 Chr.10: 132117475-132117475 on Build GRCh38
rs760645669 Chr.10: 132117445-132117445 on Build GRCh38
rs778567934 Chr.10: 132117431-132117431 on Build GRCh38

View Full Product Details