Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 2838523-2838770 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCED1A
Entrez Gene ID
64773
Gene Name
PC-esterase domain containing 1A
Gene Aliases
C20orf81, FAM113A, bA12M19.1
UniGene
Hs.29341
Gene Chromosome Location
Chr.20: 2835314-2841243 on Build GRCh38
Transcripts
NM_001271168.1 NM_022760.4 XM_005260804.2 XM_005260805.2 XM_005260806.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6092936 c.572C>T p.T191I Chr.20: 2838618-2838618 on Build GRCh38
140856 c.496G>T p.V166L Chr.20: 2838694-2838694 on Build GRCh38
6741316 c.472C>T p.R158W Chr.20: 2838718-2838718 on Build GRCh38
1681501 c.548C>T p.A183V Chr.20: 2838642-2838642 on Build GRCh38
5039119 c.546G>T p.M182I Chr.20: 2838644-2838644 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374829387 Chr.20: 2838632-2838632 on Build GRCh38
rs765474630 Chr.20: 2838704-2838704 on Build GRCh38
rs746502557 Chr.20: 2838641-2838641 on Build GRCh38
rs370121007 Chr.20: 2838719-2838719 on Build GRCh38
rs766833156 Chr.20: 2838624-2838625 on Build GRCh38
rs760885500 Chr.20: 2838583-2838583 on Build GRCh38
rs752995978 Chr.20: 2838547-2838547 on Build GRCh38
rs1016879001 Chr.20: 2838659-2838659 on Build GRCh38
rs776659286 Chr.20: 2838702-2838702 on Build GRCh38
rs746621360 Chr.20: 2838553-2838553 on Build GRCh38
rs759804117 Chr.20: 2838703-2838703 on Build GRCh38
rs769417887 Chr.20: 2838670-2838670 on Build GRCh38
rs755283839 Chr.20: 2838624-2838624 on Build GRCh38
rs775361558 Chr.20: 2838679-2838679 on Build GRCh38
rs148053451 Chr.20: 2838677-2838677 on Build GRCh38
rs766478846 Chr.20: 2838597-2838597 on Build GRCh38
rs754231866 Chr.20: 2838626-2838626 on Build GRCh38
rs770594878 Chr.20: 2838554-2838554 on Build GRCh38
rs767640037 Chr.20: 2838578-2838578 on Build GRCh38
rs768737895 Chr.20: 2838690-2838690 on Build GRCh38
rs367793028 Chr.20: 2838561-2838561 on Build GRCh38
rs764556327 Chr.20: 2838728-2838728 on Build GRCh38
rs777213218 Chr.20: 2838637-2838637 on Build GRCh38
rs754155776 Chr.20: 2838601-2838601 on Build GRCh38
rs541452774 Chr.20: 2838743-2838743 on Build GRCh38
rs144950307 Chr.20: 2838665-2838665 on Build GRCh38
rs371738728 Chr.20: 2838568-2838568 on Build GRCh38
rs747670759 Chr.20: 2838543-2838543 on Build GRCh38
rs960437637 Chr.20: 2838730-2838730 on Build GRCh38
rs946803665 Chr.20: 2838613-2838613 on Build GRCh38
rs760832333 Chr.20: 2838699-2838699 on Build GRCh38
rs752130735 Chr.20: 2838739-2838739 on Build GRCh38
rs769701416 Chr.20: 2838566-2838566 on Build GRCh38
rs1005861724 Chr.20: 2838603-2838603 on Build GRCh38
rs780766874 Chr.20: 2838749-2838749 on Build GRCh38
rs777266201 Chr.20: 2838549-2838549 on Build GRCh38
rs758932582 Chr.20: 2838635-2838635 on Build GRCh38
rs749199920 Chr.20: 2838688-2838688 on Build GRCh38
rs756803313 Chr.20: 2838647-2838647 on Build GRCh38
rs780706983 Chr.20: 2838660-2838660 on Build GRCh38
rs1055073472 Chr.20: 2838571-2838571 on Build GRCh38
rs773459319 Chr.20: 2838582-2838582 on Build GRCh38
rs577382498 Chr.20: 2838701-2838701 on Build GRCh38
rs755520684 Chr.20: 2838634-2838634 on Build GRCh38
rs762999034 Chr.20: 2838572-2838572 on Build GRCh38
rs753079343 Chr.20: 2838705-2838705 on Build GRCh38
rs765644056 Chr.20: 2838631-2838631 on Build GRCh38
rs745596454 Chr.20: 2838667-2838667 on Build GRCh38

View Full Product Details