Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 117146056-117146315 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO21
Entrez Gene ID
23014
Gene Name
F-box protein 21
Gene Aliases
FBX21
UniGene
Hs.740376
Gene Chromosome Location
Chr.12: 117143473-117190500 on Build GRCh38
Transcripts
NM_015002.2 NM_033624.2 XM_017019037.1 XM_017019038.1 XM_017019039.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6285286 c.1812T>A p.Y604* Chr.12: 117146162-117146162 on Build GRCh38
4472921 c.1814C>T p.P605L Chr.12: 117146160-117146160 on Build GRCh38
6317109 c.1786C>A p.P596T Chr.12: 117146188-117146188 on Build GRCh38
1298964 c.1825G>C p.E609Q Chr.12: 117146149-117146149 on Build GRCh38
4419086 c.1713C>T p.N571N Chr.12: 117146261-117146261 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201541949 Chr.12: 117146078-117146078 on Build GRCh38
rs779512337 Chr.12: 117146133-117146133 on Build GRCh38
rs781226460 Chr.12: 117146167-117146167 on Build GRCh38
rs773253554 Chr.12: 117146182-117146182 on Build GRCh38
rs780646873 Chr.12: 117146291-117146291 on Build GRCh38
rs752258546 Chr.12: 117146246-117146246 on Build GRCh38
rs768635405 Chr.12: 117146082-117146082 on Build GRCh38
rs1010829329 Chr.12: 117146114-117146114 on Build GRCh38
rs368180409 Chr.12: 117146097-117146097 on Build GRCh38
rs376841463 Chr.12: 117146098-117146098 on Build GRCh38
rs78022921 Chr.12: 117146180-117146180 on Build GRCh38
rs762493670 Chr.12: 117146096-117146096 on Build GRCh38
rs770496734 Chr.12: 117146192-117146192 on Build GRCh38
rs537742336 Chr.12: 117146174-117146174 on Build GRCh38
rs375044366 Chr.12: 117146231-117146231 on Build GRCh38
rs754251748 Chr.12: 117146105-117146105 on Build GRCh38
rs776485711 Chr.12: 117146083-117146083 on Build GRCh38
rs757521947 Chr.12: 117146132-117146132 on Build GRCh38
rs372082361 Chr.12: 117146224-117146224 on Build GRCh38
rs905060901 Chr.12: 117146173-117146173 on Build GRCh38
rs967545323 Chr.12: 117146095-117146095 on Build GRCh38
rs762203194 Chr.12: 117146165-117146165 on Build GRCh38
rs1015268283 Chr.12: 117146235-117146235 on Build GRCh38
rs772985333 Chr.12: 117146094-117146094 on Build GRCh38
rs3741467 Chr.12: 117146261-117146261 on Build GRCh38
rs144893839 Chr.12: 117146139-117146139 on Build GRCh38
rs919845463 Chr.12: 117146205-117146205 on Build GRCh38
rs139082982 Chr.12: 117146101-117146101 on Build GRCh38
rs769009614 Chr.12: 117146166-117146166 on Build GRCh38
rs762285013 Chr.12: 117146247-117146247 on Build GRCh38
rs368903862 Chr.12: 117146260-117146260 on Build GRCh38
rs878968748 Chr.12: 117146156-117146156 on Build GRCh38
rs780274977 Chr.12: 117146150-117146150 on Build GRCh38
rs17854201 Chr.12: 117146213-117146213 on Build GRCh38
rs375022889 Chr.12: 117146088-117146088 on Build GRCh38
rs758989840 Chr.12: 117146230-117146230 on Build GRCh38
rs143961397 Chr.12: 117146102-117146102 on Build GRCh38
rs747014658 Chr.12: 117146157-117146157 on Build GRCh38
rs769796710 Chr.12: 117146093-117146093 on Build GRCh38
rs762865500 Chr.12: 117146183-117146183 on Build GRCh38
rs148630437 Chr.12: 117146152-117146152 on Build GRCh38
rs758974861 Chr.12: 117146284-117146284 on Build GRCh38
rs770998265 Chr.12: 117146123-117146124 on Build GRCh38

View Full Product Details