Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 20618802-20619073 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HS1BP3
Entrez Gene ID
64342
Gene Name
HCLS1 binding protein 3
Gene Aliases
ETM2, HS1-BP3
UniGene
Hs.531785
Gene Chromosome Location
Chr.2: 20553861-20651661 on Build GRCh38
Transcripts
NM_022460.3 XM_017004696.1 XM_017004697.1 XM_017004698.1 XM_017004699.1 XM_017004700.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3732012 c.1168A>G p.S390G Chr.2: 20618998-20618998 on Build GRCh38
4750937 c.1111G>A p.E371K Chr.2: 20619055-20619055 on Build GRCh38
1254421 c.1167C>T p.P389P Chr.2: 20618999-20618999 on Build GRCh38
6892070 c.1165C>T p.P389S Chr.2: 20619001-20619001 on Build GRCh38
3757916 c.1162G>A p.A388T Chr.2: 20619004-20619004 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs570478819 Chr.2: 20618875-20618875 on Build GRCh38
rs556089435 Chr.2: 20618917-20618917 on Build GRCh38
rs376644274 Chr.2: 20618938-20618938 on Build GRCh38
rs200640759 Chr.2: 20618969-20618969 on Build GRCh38
rs778390412 Chr.2: 20618993-20618993 on Build GRCh38
rs866360412 Chr.2: 20618894-20618894 on Build GRCh38
rs568815381 Chr.2: 20618822-20618822 on Build GRCh38
rs1012383393 Chr.2: 20619016-20619016 on Build GRCh38
rs766534301 Chr.2: 20618953-20618953 on Build GRCh38
rs756747321 Chr.2: 20618979-20618979 on Build GRCh38
rs755306007 Chr.2: 20618963-20618963 on Build GRCh38
rs367812687 Chr.2: 20618949-20618949 on Build GRCh38
rs3732149 Chr.2: 20619004-20619004 on Build GRCh38
rs902212780 Chr.2: 20618985-20618985 on Build GRCh38
rs974155817 Chr.2: 20619051-20619051 on Build GRCh38
rs35295842 Chr.2: 20618886-20618886 on Build GRCh38
rs775987515 Chr.2: 20619041-20619041 on Build GRCh38
rs999780056 Chr.2: 20618995-20618995 on Build GRCh38
rs763000106 Chr.2: 20618944-20618944 on Build GRCh38
rs555239372 Chr.2: 20619021-20619021 on Build GRCh38
rs1043387560 Chr.2: 20618965-20618965 on Build GRCh38
rs924744271 Chr.2: 20618889-20618889 on Build GRCh38
rs1048267327 Chr.2: 20619024-20619024 on Build GRCh38
rs548018283 Chr.2: 20618865-20618865 on Build GRCh38
rs200525761 Chr.2: 20619005-20619005 on Build GRCh38
rs751768702 Chr.2: 20618958-20618958 on Build GRCh38
rs900273626 Chr.2: 20618935-20618935 on Build GRCh38
rs775330743 Chr.2: 20619022-20619022 on Build GRCh38
rs539179783 Chr.2: 20618895-20618895 on Build GRCh38
rs939743128 Chr.2: 20619045-20619045 on Build GRCh38
rs772932859 Chr.2: 20618937-20618937 on Build GRCh38
rs58985716 Chr.2: 20618904-20618904 on Build GRCh38
rs750543470 Chr.2: 20618939-20618939 on Build GRCh38
rs774972793 Chr.2: 20618963-20618963 on Build GRCh38
rs146436769 Chr.2: 20619023-20619023 on Build GRCh38
rs775828751 Chr.2: 20618947-20618948 on Build GRCh38
rs753102297 Chr.2: 20618972-20618972 on Build GRCh38
rs748854148 Chr.2: 20618996-20618996 on Build GRCh38
rs576265957 Chr.2: 20618826-20618826 on Build GRCh38

View Full Product Details