Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 32910286-32910545 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGB1
Entrez Gene ID
3688
Gene Name
integrin subunit beta 1
Gene Aliases
CD29, FNRB, GPIIA, MDF2, MSK12, VLA-BETA, VLAB
UniGene
Hs.643813
Gene Chromosome Location
Chr.10: 32900318-32958365 on Build GRCh38
Transcripts
NM_002211.3 NM_033668.2 NM_133376.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1255043 c.1975G>C p.D659H Chr.10: 32910412-32910412 on Build GRCh38
465613 c.2000A>G p.Y667C Chr.10: 32910387-32910387 on Build GRCh38
1239164 c.2018T>C p.V673A Chr.10: 32910369-32910369 on Build GRCh38
465615 c.1985C>T p.T662I Chr.10: 32910402-32910402 on Build GRCh38
684459 c.2092G>A p.D698N Chr.10: 32910295-32910295 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs61757100 Chr.10: 32910335-32910335 on Build GRCh38
rs201866476 Chr.10: 32910369-32910369 on Build GRCh38
rs375572295 Chr.10: 32910433-32910433 on Build GRCh38
rs756589020 Chr.10: 32910423-32910423 on Build GRCh38
rs768272306 Chr.10: 32910372-32910372 on Build GRCh38
rs776347556 Chr.10: 32910379-32910379 on Build GRCh38
rs202067892 Chr.10: 32910441-32910441 on Build GRCh38
rs878933017 Chr.10: 32910464-32910464 on Build GRCh38
rs371061707 Chr.10: 32910478-32910478 on Build GRCh38
rs201149175 Chr.10: 32910489-32910489 on Build GRCh38
rs748756265 Chr.10: 32910515-32910516 on Build GRCh38
rs1019998537 Chr.10: 32910430-32910430 on Build GRCh38
rs150610387 Chr.10: 32910368-32910368 on Build GRCh38
rs778272545 Chr.10: 32910432-32910432 on Build GRCh38
rs769816472 Chr.10: 32910504-32910504 on Build GRCh38
rs975469189 Chr.10: 32910502-32910502 on Build GRCh38
rs866514558 Chr.10: 32910307-32910308 on Build GRCh38
rs559086752 Chr.10: 32910506-32910506 on Build GRCh38
rs761432412 Chr.10: 32910494-32910494 on Build GRCh38
rs201154961 Chr.10: 32910309-32910309 on Build GRCh38
rs965493360 Chr.10: 32910491-32910493 on Build GRCh38
rs751742962 Chr.10: 32910410-32910410 on Build GRCh38
rs766555231 Chr.10: 32910407-32910407 on Build GRCh38
rs202236577 Chr.10: 32910347-32910347 on Build GRCh38
rs762685966 Chr.10: 32910403-32910403 on Build GRCh38
rs199935806 Chr.10: 32910360-32910360 on Build GRCh38
rs770349356 Chr.10: 32910320-32910320 on Build GRCh38
rs761812073 Chr.10: 32910380-32910380 on Build GRCh38
rs746641897 Chr.10: 32910481-32910481 on Build GRCh38
rs750195307 Chr.10: 32910394-32910394 on Build GRCh38
rs768464221 Chr.10: 32910486-32910486 on Build GRCh38
rs200928291 Chr.10: 32910416-32910416 on Build GRCh38
rs141385192 Chr.10: 32910448-32910448 on Build GRCh38
rs752811283 Chr.10: 32910418-32910418 on Build GRCh38
rs765315222 Chr.10: 32910390-32910390 on Build GRCh38
rs139673287 Chr.10: 32910414-32910414 on Build GRCh38
rs555032553 Chr.10: 32910317-32910317 on Build GRCh38
rs200905323 Chr.10: 32910341-32910341 on Build GRCh38
rs201846400 Chr.10: 32910350-32910350 on Build GRCh38

View Full Product Details