Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 25549199-25549464 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LMNTD1
Entrez Gene ID
160492
Gene Name
lamin tail domain containing 1
Gene Aliases
IFLTD1, PAS1C1
UniGene
Hs.44647
Gene Chromosome Location
Chr.12: 25476082-25648604 on Build GRCh38
Transcripts
NM_001145727.2 NM_001145728.2 NM_001145729.1 NM_001256266.1 NM_152590.3 XM_011520575.1 XM_011520576.2 XM_011520577.2 XM_011520578.2 XM_011520579.2 XM_011520580.1 XM_011520582.1 XM_011520583.1 XM_011520584.1 XM_011520585.2 XM_017018891.1 XM_017018892.1 XM_017018893.1 XM_017018894.1 XM_017018895.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5960616 c.233G>C p.R78T Chr.12: 25549403-25549403 on Build GRCh38
7264807 c.196C>T p.P66S Chr.12: 25549440-25549440 on Build GRCh38
7332479 c.248C>A p.T83K Chr.12: 25549388-25549388 on Build GRCh38
3986826 c.218G>T p.G73V Chr.12: 25549355-25549355 on Build GRCh38
3459523 c.253G>A p.G85R Chr.12: 25549383-25549383 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1038560505 Chr.12: 25549261-25549261 on Build GRCh38
rs779637082 Chr.12: 25549413-25549413 on Build GRCh38
rs773906819 Chr.12: 25549359-25549359 on Build GRCh38
rs758092258 Chr.12: 25549279-25549279 on Build GRCh38
rs753961773 Chr.12: 25549376-25549376 on Build GRCh38
rs777570456 Chr.12: 25549293-25549293 on Build GRCh38
rs762335329 Chr.12: 25549346-25549346 on Build GRCh38
rs376996046 Chr.12: 25549416-25549416 on Build GRCh38
rs755401608 Chr.12: 25549410-25549410 on Build GRCh38
rs779539410 Chr.12: 25549323-25549323 on Build GRCh38
rs540331705 Chr.12: 25549262-25549262 on Build GRCh38
rs933916054 Chr.12: 25549227-25549227 on Build GRCh38
rs745353522 Chr.12: 25549311-25549311 on Build GRCh38
rs140866724 Chr.12: 25549256-25549256 on Build GRCh38
rs772695737 Chr.12: 25549352-25549352 on Build GRCh38
rs145363780 Chr.12: 25549249-25549249 on Build GRCh38
rs750414068 Chr.12: 25549408-25549408 on Build GRCh38
rs751346910 Chr.12: 25549395-25549395 on Build GRCh38
rs952776576 Chr.12: 25549305-25549305 on Build GRCh38
rs749001637 Chr.12: 25549329-25549329 on Build GRCh38
rs768419387 Chr.12: 25549437-25549437 on Build GRCh38
rs958376259 Chr.12: 25549378-25549378 on Build GRCh38
rs367788487 Chr.12: 25549314-25549314 on Build GRCh38
rs369352018 Chr.12: 25549439-25549439 on Build GRCh38
rs74713971 Chr.12: 25549270-25549270 on Build GRCh38
rs753785728 Chr.12: 25549357-25549358 on Build GRCh38
rs561666746 Chr.12: 25549265-25549265 on Build GRCh38
rs544482469 Chr.12: 25549308-25549308 on Build GRCh38
rs768853560 Chr.12: 25549333-25549333 on Build GRCh38
rs781082064 Chr.12: 25549396-25549396 on Build GRCh38
rs374562785 Chr.12: 25549272-25549272 on Build GRCh38
rs185106362 Chr.12: 25549252-25549252 on Build GRCh38
rs760643100 Chr.12: 25549363-25549363 on Build GRCh38
rs766514233 Chr.12: 25549371-25549371 on Build GRCh38
rs751436937 Chr.12: 25549306-25549306 on Build GRCh38
rs763807179 Chr.12: 25549393-25549393 on Build GRCh38
rs533279790 Chr.12: 25549341-25549341 on Build GRCh38
rs377307789 Chr.12: 25549391-25549391 on Build GRCh38

View Full Product Details