Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 48514243-48514468 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PORCN
Entrez Gene ID
64840
Gene Name
porcupine homolog (Drosophila)
Gene Aliases
DHOF, FODH, MG61, PORC, PPN
UniGene
Hs.386453
Gene Chromosome Location
Chr.X: 48508954-48520814 on Build GRCh38
Transcripts
NM_001282167.1 NM_022825.3 NM_203473.2 NM_203474.1 NM_203475.2 XM_005272635.2 XM_005272636.2 XM_005272637.1 XM_006724545.3 XM_006724546.3 XM_006724547.1 XM_011543948.2 XM_017029735.1 XM_017029736.1

SNP Detail

SNP ID Chromosome Location
rs782496390 Chr.X: 48514342-48514342 on Build GRCh38
rs782730333 Chr.X: 48514332-48514332 on Build GRCh38
rs369672830 Chr.X: 48514325-48514325 on Build GRCh38
rs781876155 Chr.X: 48514374-48514374 on Build GRCh38
rs782663149 Chr.X: 48514264-48514264 on Build GRCh38
rs376623776 Chr.X: 48514321-48514321 on Build GRCh38
rs782175760 Chr.X: 48514309-48514309 on Build GRCh38
rs782751723 Chr.X: 48514320-48514320 on Build GRCh38
rs782508840 Chr.X: 48514408-48514408 on Build GRCh38
rs370092224 Chr.X: 48514301-48514301 on Build GRCh38
rs781791638 Chr.X: 48514333-48514333 on Build GRCh38
rs899748308 Chr.X: 48514331-48514331 on Build GRCh38
rs781949383 Chr.X: 48514315-48514315 on Build GRCh38
rs782652383 Chr.X: 48514413-48514413 on Build GRCh38
rs995793394 Chr.X: 48514418-48514418 on Build GRCh38
rs782256540 Chr.X: 48514367-48514368 on Build GRCh38
rs782285872 Chr.X: 48514269-48514269 on Build GRCh38
rs782735906 Chr.X: 48514343-48514343 on Build GRCh38
rs782131250 Chr.X: 48514317-48514317 on Build GRCh38
rs781928782 Chr.X: 48514306-48514306 on Build GRCh38
rs782462957 Chr.X: 48514368-48514368 on Build GRCh38
rs200707168 Chr.X: 48514314-48514314 on Build GRCh38
rs782673953 Chr.X: 48514378-48514378 on Build GRCh38

View Full Product Details