Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 190437362-190437619 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MFSD6
Entrez Gene ID
54842
Gene Name
major facilitator superfamily domain containing 6
Gene Aliases
MMR2, hMMR2
UniGene
Hs.418581
Gene Chromosome Location
Chr.2: 190408355-190502315 on Build GRCh38
Transcripts
NM_017694.3 XM_005246654.2 XM_005246656.3 XM_011511368.1 XM_017004392.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6258851 c.1353G>T p.L451L Chr.2: 190437382-190437382 on Build GRCh38
3575585 c.1443C>T p.L481L Chr.2: 190437472-190437472 on Build GRCh38
5555562 c.1372G>T p.G458C Chr.2: 190437401-190437401 on Build GRCh38
221978 c.1457C>T p.S486L Chr.2: 190437486-190437486 on Build GRCh38
4956115 c.1345T>G p.S449A Chr.2: 190437374-190437374 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs927027314 Chr.2: 190437514-190437514 on Build GRCh38
rs373172725 Chr.2: 190437587-190437587 on Build GRCh38
rs759762569 Chr.2: 190437499-190437499 on Build GRCh38
rs748962411 Chr.2: 190437433-190437433 on Build GRCh38
rs111906485 Chr.2: 190437550-190437550 on Build GRCh38
rs1046390610 Chr.2: 190437481-190437481 on Build GRCh38
rs755625619 Chr.2: 190437578-190437578 on Build GRCh38
rs774315733 Chr.2: 190437456-190437456 on Build GRCh38
rs780520943 Chr.2: 190437393-190437393 on Build GRCh38
rs779837253 Chr.2: 190437419-190437419 on Build GRCh38
rs369036382 Chr.2: 190437493-190437493 on Build GRCh38
rs372452117 Chr.2: 190437496-190437496 on Build GRCh38
rs765546841 Chr.2: 190437538-190437538 on Build GRCh38
rs758976051 Chr.2: 190437392-190437392 on Build GRCh38
rs529962340 Chr.2: 190437488-190437488 on Build GRCh38
rs539999264 Chr.2: 190437454-190437454 on Build GRCh38
rs761602994 Chr.2: 190437472-190437472 on Build GRCh38
rs752362114 Chr.2: 190437577-190437578 on Build GRCh38
rs938463240 Chr.2: 190437521-190437521 on Build GRCh38
rs952177768 Chr.2: 190437397-190437397 on Build GRCh38
rs377702122 Chr.2: 190437580-190437580 on Build GRCh38
rs920677182 Chr.2: 190437384-190437384 on Build GRCh38
rs374771250 Chr.2: 190437479-190437479 on Build GRCh38
rs937487897 Chr.2: 190437553-190437553 on Build GRCh38
rs745362203 Chr.2: 190437403-190437403 on Build GRCh38
rs34551832 Chr.2: 190437402-190437403 on Build GRCh38
rs772022225 Chr.2: 190437478-190437478 on Build GRCh38
rs34578589 Chr.2: 190437417-190437418 on Build GRCh38
rs144388154 Chr.2: 190437417-190437417 on Build GRCh38
rs764632947 Chr.2: 190437573-190437573 on Build GRCh38
rs181807426 Chr.2: 190437577-190437577 on Build GRCh38

View Full Product Details