Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 2589853-2590102 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NDC80
Entrez Gene ID
10403
Gene Name
NDC80, kinetochore complex component
Gene Aliases
HEC, HEC1, HsHec1, KNTC2, TID3, hsNDC80
UniGene
Hs.414407
Gene Chromosome Location
Chr.18: 2571511-2616635 on Build GRCh38
Transcripts
NM_006101.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
987364 c.884C>T p.S295L Chr.18: 2590031-2590031 on Build GRCh38
6839607 c.914G>A p.G305E Chr.18: 2590061-2590061 on Build GRCh38
3524724 c.906C>T p.S302S Chr.18: 2590053-2590053 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs144899651 Chr.18: 2590032-2590032 on Build GRCh38
rs995699897 Chr.18: 2589996-2589996 on Build GRCh38
rs8097280 Chr.18: 2589930-2589930 on Build GRCh38
rs757145108 Chr.18: 2589953-2589953 on Build GRCh38
rs773351560 Chr.18: 2590005-2590005 on Build GRCh38
rs767815895 Chr.18: 2589999-2589999 on Build GRCh38
rs539828123 Chr.18: 2590052-2590052 on Build GRCh38
rs116295956 Chr.18: 2589898-2589898 on Build GRCh38
rs890060574 Chr.18: 2590058-2590058 on Build GRCh38
rs760970000 Chr.18: 2590028-2590028 on Build GRCh38
rs753954742 Chr.18: 2590046-2590046 on Build GRCh38
rs367786547 Chr.18: 2590009-2590009 on Build GRCh38
rs959054594 Chr.18: 2589950-2589950 on Build GRCh38
rs891259560 Chr.18: 2589880-2589880 on Build GRCh38
rs140253066 Chr.18: 2590047-2590047 on Build GRCh38

View Full Product Details