Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 156327958-156328195 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NR4A2
Entrez Gene ID
4929
Gene Name
nuclear receptor subfamily 4 group A member 2
Gene Aliases
HZF-3, NOT, NURR1, RNR1, TINUR
UniGene
Hs.563344
Gene Chromosome Location
Chr.2: 156324432-156334683 on Build GRCh38
Transcripts
NM_006186.3 XM_005246621.3 XM_005246622.3 XM_006712553.3 XM_011511246.1 XM_017004219.1 XM_017004220.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4304622 c.1075A>C p.K359Q Chr.2: 156327967-156327967 on Build GRCh38
1217803 c.1030C>T p.R344C Chr.2: 156327979-156327979 on Build GRCh38
4304623 c.1074G>A p.S358S Chr.2: 156327968-156327968 on Build GRCh38
4763089 c.1055G>A p.R352Q Chr.2: 156327987-156327987 on Build GRCh38
3299592 c.1041G>A p.S347S Chr.2: 156327968-156327968 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777116116 Chr.2: 156328031-156328031 on Build GRCh38
rs374052982 Chr.2: 156328028-156328028 on Build GRCh38
rs767495441 Chr.2: 156328059-156328059 on Build GRCh38
rs547013920 Chr.2: 156328115-156328115 on Build GRCh38
rs765519917 Chr.2: 156328041-156328041 on Build GRCh38
rs1021259720 Chr.2: 156328083-156328083 on Build GRCh38
rs766046906 Chr.2: 156328101-156328101 on Build GRCh38
rs772647221 Chr.2: 156327989-156327989 on Build GRCh38
rs778308914 Chr.2: 156328132-156328132 on Build GRCh38
rs773451412 Chr.2: 156328052-156328052 on Build GRCh38
rs934204568 Chr.2: 156328033-156328033 on Build GRCh38
rs16840249 Chr.2: 156328030-156328030 on Build GRCh38
rs975690434 Chr.2: 156328029-156328029 on Build GRCh38
rs561438884 Chr.2: 156328084-156328084 on Build GRCh38
rs902574234 Chr.2: 156328119-156328119 on Build GRCh38
rs763068811 Chr.2: 156328054-156328054 on Build GRCh38
rs775789577 Chr.2: 156328019-156328019 on Build GRCh38
rs992323860 Chr.2: 156328163-156328163 on Build GRCh38
rs1011747848 Chr.2: 156328082-156328082 on Build GRCh38
rs1056830709 Chr.2: 156328154-156328154 on Build GRCh38
rs530100216 Chr.2: 156328092-156328092 on Build GRCh38
rs946892225 Chr.2: 156328104-156328104 on Build GRCh38
rs551170575 Chr.2: 156328073-156328073 on Build GRCh38
rs1031245505 Chr.2: 156328133-156328133 on Build GRCh38
rs967464857 Chr.2: 156328085-156328085 on Build GRCh38
rs961675956 Chr.2: 156328025-156328025 on Build GRCh38
rs762169789 Chr.2: 156328035-156328035 on Build GRCh38
rs760687343 Chr.2: 156328100-156328100 on Build GRCh38

View Full Product Details